Skip to Content
Merck
All Photos(1)

Key Documents

HPA018284

Sigma-Aldrich

Anti-GGCX antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Gamma-glutamyl carboxylase, Anti-Vitamin K gamma glutamyl carboxylase, Anti-Vitamin K-dependent gamma-carboxylase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

FPGLHLENFVSEDLGNTSIQLLQGEVTVELVAEQKNQTLREGEKMQLPAGEYHKVYTTSPSPSCYMYVYVNTTELALEQDLAYLQELKEKVENGSETGPLPPELQPLLEGEVK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GGCX(2677)

General description

The gene GGCX (γ-glutamyl carboxylase) is mapped to human chromosome 2p12. The protein is localized in the endoplasmic reticulum (ER) and the Golgi complex. The amino-terminus of the protein is present in the cytoplasm and the carboxyl-terminus in the lumen of the ER/Golgi.

Immunogen

Vitamin K-dependent gamma-carboxylase recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

GGCX (γ-glutamyl carboxylase) carboxylates glutamate residues located in the γ-carboxyglutamic domain of vitamin K-dependent coagulation factors. Missense mutations in GGCX are associated with deficiency of the vitamin K-dependent coagulation factors, a rare autosomal recessive inherited bleeding disorder. Similarly, mutations in GGCX are associated with pseudoxanthoma elasticum like phenotype, characterized with excessive skin folding and a deficiency of the vitamin K-dependent clotting fact. GGCX also affects the metabolism of warfarin, an anticoagulant. GGCX polymorphism affects the warfarin dose in patients with atrial fibrillation.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73230

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Olivier M Vanakker et al.
The Journal of investigative dermatology, 127(3), 581-587 (2006-11-18)
Data on six patients with a Pseudoxanthoma Elasticum (PXE)-like phenotype, characterized by excessive skin folding (resembling cutis laxa) and a deficiency of the vitamin K-dependent clotting factors (II, VII, IX, and X) are presented. A comparison is made between the
Xiayizha Kamali et al.
Lipids in health and disease, 12, 149-149 (2013-10-24)
To study the effects of γ-glutamyl carboxylase (GGCX) rs2592551 polymorphism on warfarin dose in atrial fibrillation patients in Xinjiang region. Polymerase chain reaction - restriction fragment length polymorphism and direct sequencing methods were used to detect the rs2592551 genotype in
Yundan Liang et al.
Genetic testing and molecular biomarkers, 17(12), 932-936 (2013-08-15)
Warfarin is a commonly used anticoagulant with a narrow therapeutic range and large interindividual differences in dosing requirements. Previously, studies have identified that the interindividual variability was influenced by varieties of factors, including age, body size, vitamin K intake, interacting
Simone Rost et al.
British journal of haematology, 126(4), 546-549 (2004-08-04)
Hereditary combined deficiency of the vitamin K-dependent coagulation factors II, VII, IX, X, protein C, S and protein Z (VKCFD) is a very rare autosomal recessive inherited bleeding disorder. The phenotype may result from functional deficiency of either the gamma-glutamyl
B Brenner et al.
Blood, 92(12), 4554-4559 (1998-12-09)
To identify potential mutations in the gamma-glutamyl carboxylase gene, the sequence of all exons and intron/exon borders was determined in 4 patients from a consanguineous kindred with combined deficiency of all vitamin K-dependent procoagulants and anticoagulants and results were compared

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service