Skip to Content
Merck
All Photos(1)

Key Documents

SHC002

Sigma-Aldrich

MISSION® pLKO.1-puro Non-Mammalian shRNA Control Plasmid DNA

Targets no known mammalian genes

Synonym(s):

MISSION® Control Vectors

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41106609
NACRES:
NA.51

Quality Level

product line

MISSION®

concentration

500 ng/μL in TE buffer; DNA (10μg of plasmid DNA)

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

MISSION® pLKO.1-puro non-mammalian shRNA control plasmid DNA has been used:
  • to transduce RLE-6TN cells
  • for the validation of Oct4-green fluorescent protein (GFP) mouse embryonic fibroblasts (MEFs)
  • as a negative control for RNA knockdown

MISSION®pLKO.1-puro Non-Mammalian shRNA Control Plasmid DNA has been used:
  • for the knockdown of transmembrane prostate androgen-induced RNA (TMEPAI)
  • in the infection mixture for lentivirus preparation
  • for generating the puromycin-resistant ST14A (rat striatal neuronal cell line) cell populations
  • to induce Ago2 (argonaute-2) down-regulation

Legal Information

Use of this product is subject to one or more license agreements. For details, please see http://sigmaaldrich.com/missionlicense.
MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification of a gene expression signature associated with the metastasis suppressor function of NME1: prognostic value in human melanoma
Leonard MK, et al.
Laboratory Investigation; a Journal of Technical Methods and Pathology, 98(3), 327-327 (2018)
TMEPAI/PMEPA 1 enhances tumorigenic activities in lung cancer cells
Vo Nguyen TT, et al.
Cancer Science, 105(3), 334-341 (2014)
JNK1 Deficient Insulin-Producing Cells Are Protected against Interleukin-1?-Induced Apoptosis Associated with Abrogated Myc Expression.
Prause M
Journal of Diabetes Research (2016)
SFPQ?NONO and XLF function separately and together to promote DNA double-strand break repair via canonical nonhomologous end joining.
Jaafar l
Nucleic Acids Research, 45(4), 1848-1859 (2017)
Platelet-independent adhesion of calcium-loaded erythrocytes to von Willebrand factor.
Smeets MW
PLoS ONE, 12.3 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service