Skip to Content
Merck
All Photos(2)

Documents

HPA010778

Sigma-Aldrich

Anti-FAM163A antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-C1orf76 antibody produced in rabbit, Anti-Neuroblastoma-derived secretory protein antibody produced in rabbit, Anti-UPF0417 protein C1orf76 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

YYCCKKSGTEVADEEEEREHDLPTHPRGPTCNACSSQALDGRGSLAPLTSEPCSQPCGVAASHCTTCSPYSSPFYIRTADMVPNGGGGERLSFAPTYYKEGGPPSLKLAAPQSYPVTWPGSGREAFTNPRAIST

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

FAM163A (family with sequence similarity 163, member A) is a secreted protein, which is also called neuroblastoma-derived secretory protein (NDSP). This gene is located on human chromosome 1q25.2. The encoded protein contains a C-C motif and a signal peptide made of 30 amino acids. It consists of 167 amino acids, and is secreted as both, a monomer and a homodimer. Its molecular weight is 18kDa. It is normally expressed in adrenal gland, brain, thymus, colon, ileum, uterus, and testes, where it has the highest level of expression in testes.

Immunogen

UPF0417 protein C1orf76 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

FAM163A (family with sequence similarity 163, member A) is up-regulated in neuroblastoma, as opposed to normal tissues and adult and pediatric cancers. It therefore, plays a role in tumorigenesis of neuroblastoma. This might be achieved by it acting as a ligand for growth factors receptors. It prevents anoikis and supports anchorage independent growth, via ERK (extracellular signal-regulated kinases)-mediated proliferation pathway. FAM163A can therefore, be used as therapeutic target in neuroblastoma. Its mRNA levels also correlate with the risk level in neuroblastoma, and thus, have potential as a marker to determine the metastasis in bone marrow.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72031

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sanjeev A Vasudevan et al.
Molecular cancer therapeutics, 8(8), 2478-2489 (2009-08-13)
Secreted proteins such as growth factors, cytokines, and chemokines play important roles in tumor development. Through expression microarray and bioinformatic analysis, we discovered a novel secreted protein, neuroblastoma-derived secretory protein (NDSP). The NDSP gene is found on chromosome 1q25.2 and
Sanjeev A Vasudevan et al.
Journal of pediatric surgery, 42(1), 148-152 (2007-01-09)
In advanced-stage neuroblastoma, bulky disease and systemic dissemination can be controlled with intense surgical and medical therapies; however, recurrence rates are very high in this group indicating that residual disease is rarely eradicated. The need to detect residual disease and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service