Skip to Content
Merck
All Photos(6)

Key Documents

HPA008763

Sigma-Aldrich

Anti-GRN antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Granulins precursor, Anti-PEPI, Anti-Proepithelin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

rat, mouse, human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

DSQFECPDFSTCCVMVDGSWGCCPMPQASCCEDRVHCCPHGAFCDLVHTRCITPTGTHPLAKKLPAQRTNRAVALSSSVMCPDARSRCPDGSTCCELPSGKYGCCPMPNATCCSDHLHCCPQDTVCDLIQSKCLSKENATTDLL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GRN(2896)

General description

GRN (granulin) is a large cysteine rich glycoprotein, also called progranulin, which has a molecular weight of 88,000kDa. It is the largest member of granulin/epithelin family of growth factors. This family is characterized a unique double cysteine rich motif.

Immunogen

Granulins precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-GRN antibody has been used in:
  • granulin immunohistochemistry
  • immunoblotting
  • tissue microarrays

Anti-GRN antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

GRN (granulin) is a growth and survival factor, which is involved in drug resistance and cancer. It is up-regulated in breast cancer, where it is responsible for tumorigenesis, drug resistance, invasion, metastasis and angiogenesis. It is up-regulated in non-small cell lung cancer (NSCLC), and it has potential as a prognostic marker and therapeutic target in the same. Serum levels of this protein correlate with visceral fat and increased level of liver enzymes. Thus, serum granulin level has potential as a marker for liver dysfunction.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86882

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Malene Overby et al.
The Journal of biological chemistry, 299(12), 105446-105446 (2023-11-11)
Increasing evidence suggests that aberrant regulation of sortilin ectodomain shedding can contribute to amyloid-β pathology and frontotemporal dementia, although the mechanism by which this occurs has not been elucidated. Here, we probed for novel binding partners of sortilin using multiple
Songkiad Upontain et al.
Asian Pacific journal of cancer prevention : APJCP, 19(9), 2437-2445 (2018-09-27)
The secreted growth factor granulin (GRN) is upregulated during diverse epithelial cancers. GRN stimulates cell growth and development while inhibiting apoptosis. Orthologues of vertebrate granulins evolved in other animals including the liver fluke Opisthorchis viverrini. Curiously, liver fluke granulin, termed
Human tumors instigate granulin-expressing hematopoietic cells that promote malignancy by activating stromal fibroblasts in mice
Elkabets M, et al.
The Journal of Clinical Investigation, 121(2), 784-799 (2011)
Yuko Tanaka et al.
Endocrine journal, 61(12), 1191-1196 (2014-09-19)
Progranulin (PRGN) was recently identified as one of the adipokines involved in the development of insulin resistance. Thus, the aim of this study was to explore the importance of PRGN as a novel marker for metabolic diseases in Japanese. A
Murine knockin model for progranulin-deficient frontotemporal dementia with nonsense-mediated mRNA decay
Nguyen A D, et al.
Proceedings of the National Academy of Sciences of the USA, 115(12), E2849-E2858 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service