Skip to Content
Merck
All Photos(5)

Key Documents

HPA008455

Sigma-Aldrich

Anti-MCL1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Bcl-2-related protein EAT/mcl1 antibody produced in rabbit, Anti-Induced myeloid leukemia cell differentiation protein Mcl-1 antibody produced in rabbit, Anti-mcl1/EAT antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

DAIMSPEEELDGYEPEPLGKRPAVLPLLELVGESGNNTSTDGSLPSTPPPAEEEEDELYRQSLEIISRYLREQATGAKDTKPMGRSGATSRKALETLRRVGDG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MCL1(4170)

General description

Induced myeloid leukemia cell differentiation protein Mcl-1(MCL1) belongs to the Bcl-2 family and contains the characteristic Bcl-2 homology (BH) domain. The gene encoding this protein is present on chromosome 1q21.

Immunogen

myeloid cell leukemia 1

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunocytochemistry (1 paper)
Immunohistochemistry (1 paper)

Biochem/physiol Actions

Induced myeloid leukemia cell differentiation protein Mcl-1(MCL1) plays an important role in inhibiting apoptosis and is expressed only during particular stages of cell differentiation and in response to specific signals. It is a viability-promoting protein that interacts with fortilin (an anti-apoptotic protein) and regulates cell viability. Specific differentiation- and apoptosis-inducing stimuli enhance the expression of this protein. MCL1 acts as a chaperone of fortilin by binding and stabilizing it. It also interacts and negatively regulates the proliferating cell nuclear antigen (PCNA), which a cell cycle-regulatory protein essential for G1 to S phase transition. Hence, it functions to retard the cell cycle progression.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70113

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K J Townsend et al.
Oncogene, 17(10), 1223-1234 (1998-10-15)
Members of both the mitogen activated protein (MAP) kinase and BCL2 gene families, acting in concert with other gene products, are involved in the regulation of cell viability. However, the relationship between these families, and the signal transduction networks that
Nathalie Roders et al.
Frontiers in immunology, 9, 787-787 (2018-05-10)
B cells play a major role in the antibody-mediated rejection (AMR) of solid organ transplants, a major public health concern. The germinal center (GC) is involved in the generation of donor-specific antibody-producing plasma cells and memory B cells, which are
R W Craig et al.
Genomics, 23(2), 457-463 (1994-09-15)
The MCL1 gene, recently identified in a myeloid leukemia cell line, has sequence similarity to BCL2, the gene at the t(14;18) translocation in follicular lymphoma. The chromosomal location of MCL1 has now been determined. The human locus (MCL1) was mapped
C D Bingle et al.
The Journal of biological chemistry, 275(29), 22136-22146 (2000-04-15)
Mcl-1 is a member of the Bcl-2 family that is regulated transcriptionally and post-transcriptionally, with expression of the full-length Mcl-1-encoded gene product resulting in enhanced cell survival. As reported here, the human Mcl-1 gene can also undergo differential splicing, which
Physical and functional interaction between myeloid cell leukemia 1 protein (MCL1) and Fortilin. The potential role of MCL1 as a fortilin chaperone.
D Zhang
The Journal of Biological Chemistry, 277(40), 37430-37438 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service