Skip to Content
Merck
All Photos(10)

Key Documents

HPA023370

Sigma-Aldrich

Anti-PCM1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1

Synonym(s):

Pcm1 Antibody, Pcm1 Antibody - Anti-PCM1 antibody produced in rabbit, Anti-PCM-1, Anti-Pericentriolar material 1 protein, Anti-hPCM-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:1000-1:2500

immunogen sequence

EEEGVSGASLSSHRSSLVDEHPEDAEFEQKINRLMAAKQKLRQLQDLVAMVQDDDAAQGVISASASNLDDFYPAEEDTKQNSNNTRGNANKTQKDT

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PCM1(5108)

General description

The gene PCM1 (pericentriolar material 1) is mapped to human chromosome 8p22. The encoded protein localizes to cytoplasmic granules known as ‘centriolar satellites′, which are present around the centrosome. It has a molar mass of 228kD protein.

Immunogen

Pericentriolar material 1 protein recombinant protein epitope signature tag (PrEST)

Application

Anti-PCM1 antibody produced in rabbit has been used as a primary antibody in immunofluorescence. It has also been used for immunohistochemistry study and for flow cytometry and magnetic-assisted cell sorting assay.
All Prestige Antibodies®Powered by Atlas Antibodies is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

The gene PCM1 (pericentriolar material 1) encodes a large centrosomal protein with multiple coiled-coil domains that forms a complex with disrupted-in-schizophrenia 1 (DISC1) and Bardet-Biedl syndrome 4 protein (BBS4). This binding helps in synergistic targeting of PCM1 and other cargo proteins to the centrosome. It functions as a scaffold and facilitates the targeting of several proteins to the centrosome in a dynein motor-dependent manner and helps in regulating microtubular dynamics. The centriolar satellites that contain PCM1 protein participate in microtubule- and dynactin-dependent recruitment of proteins to the centrosome and mediate the assembly of centrosomal proteins and microtubule organization. Defects in this gene are associated with reduction in gray matter volume and susceptibility to schizophrenia.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST76187

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antoinette F van Ouwerkerk et al.
Nature communications, 10(1), 4755-4755 (2019-10-20)
Disease-associated genetic variants that lie in non-coding regions found by genome-wide association studies are thought to alter the functionality of transcription regulatory elements and target gene expression. To uncover causal genetic variants, variant regulatory elements and their target genes, here
Benjamin R Nixon et al.
JCI insight, 2(4), e90656-e90656 (2017-02-28)
It remains unclear how perturbations in cardiomyocyte sarcomere function alter postnatal heart development. We utilized murine models that allowed manipulation of cardiac myosin-binding protein C (MYBPC3) expression at critical stages of cardiac ontogeny to study the response of the postnatal
Ralf Gilsbach et al.
Nature communications, 5, 5288-5288 (2014-10-23)
The heart is a highly specialized organ with essential function for the organism throughout life. The significance of DNA methylation in shaping the phenotype of the heart remains only partially known. Here we generate and analyse DNA methylomes from highly
Noelia Muñoz-Martín et al.
Development (Cambridge, England), 146(3) (2019-01-16)
Myc is considered an essential transcription factor for heart development, but cardiac defects have only been studied in global Myc loss-of-function models. Here, we eliminated Myc by recombining a Myc floxed allele with the Nkx2.5Cre driver. We observed no anatomical
Sara Blocquiaux et al.
Experimental gerontology, 133, 110860-110860 (2020-02-06)
Ageing is associated with an attenuated hypertrophic response to resistance training and periods of training interruptions. Hence, elderly would benefit from the 'muscle memory' effects of resistance training on muscle strength and mass during detraining and retraining. As the underlying

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service