Skip to Content
Merck
All Photos(3)

Key Documents

HPA004795

Sigma-Aldrich

Anti-SLC10A2 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-ASBT, Anti-IBAT, Anti-ISBT, Anti-Ileal sodium/bile acid cotransporter

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

NKAEIPESKENGTEPESSFYKANGGFQPDE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC10A2(6555)

Immunogen

Ileal sodium/bile acid cotransporter recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

SLC10A2 (solute carrier family 10, sodium/bile acid cotransporter, member 2) is a seven transmembrane (TM) domain protein. It acts as a bile acid transporter in the presence of sodium molecules. It is highly involved in the cholesterol metabolism. In the metabolism process, it triggers the enterohepatic circulation of bile salts. Mutation in SLC10A2 is associated with primary bile acid malabsorption (PBAM), an idiopathic intestinal disorder.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86075

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sabine Middendorp et al.
Stem cells (Dayton, Ohio), 32(5), 1083-1091 (2014-02-06)
Differentiation and specialization of epithelial cells in the small intestine are regulated in two ways. First, there is differentiation along the crypt-villus axis of the intestinal stem cells into absorptive enterocytes, Paneth, goblet, tuft, enteroendocrine, or M cells, which is
P Oelkers et al.
The Journal of clinical investigation, 99(8), 1880-1887 (1997-04-15)
Primary bile acid malabsorption (PBAM) is an idiopathic intestinal disorder associated with congenital diarrhea, steatorrhea, interruption of the enterohepatic circulation of bile acids, and reduced plasma cholesterol levels. The molecular basis of PBAM is unknown, and several conflicting mechanisms have
Yongtao Xiao et al.
EBioMedicine, 35, 133-141 (2018-08-15)
Although the pathogenesis of intestinal failure (IF)-associated liver disease (IFALD) is uncertain, IF-associated cholestasis mediated by the combination of intestinal injury and parenteral nutrition (PN) can lead to disturbed hepatocyte bile acids (BA) homeostasis and cause liver damages. We here
Eric Y Zhang et al.
Biochemistry, 43(36), 11380-11392 (2004-09-08)
The apical sodium-dependent bile acid transporter (ASBT, SLC10A2) facilitates the enterohepatic circulation of bile salts and plays a key role in cholesterol metabolism. The membrane topology of ASBT was initially scanned using a consensus topography analysis that predominantly predicts a
Qianhui Yu et al.
Cell, 184(12), 3281-3298 (2021-05-22)
Organs are composed of diverse cell types that traverse transient states during organogenesis. To interrogate this diversity during human development, we generate a single-cell transcriptome atlas from multiple developing endodermal organs of the respiratory and gastrointestinal tract. We illuminate cell

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service