Skip to Content
Merck
All Photos(4)

Key Documents

HPA003239

Sigma-Aldrich

Anti-PCMT1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-L-Isoaspartyl protein carboxyl methyltransferase antibody produced in rabbit, Anti-PIMT antibody produced in rabbit, Anti-Protein L-isoaspartyl/D-aspartyl methyltransferase antibody produced in rabbit, Anti-Protein-β-aspartate methyltransferase antibody produced in rabbit, Anti-Protein-L-isoaspartate(D-aspartate) O-methyltransferase antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

rat, mouse, human

enhanced validation

RNAi knockdown
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

AKALDVGSGSGILTACFARMVGCTGKVIGIDHIKELVDDSVNNVRKDDPTLLSSGRVQLVVGDGRMGYAEEAPYDAIHVGAAAPVVPQALIDQLKPGGRLILPVGPAGGNQMLEQYDKLQDGSIKMKPLMGVIYVPLTDKEKQWS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PCMT1(5110)

General description

PCMT1 (protein-L-isoaspartate (D-aspartate) O-methyltransferase) gene encodes a protein that belongs to type II class of protein carboxyl methyltransferase enzymes. This gene is mapped to human chromosome 6q24-25.

Immunogen

Protein-L-isoaspartate(D-aspartate) O-methyltransferase recombinant protein epitope signature tag (PrEST)

Application

Anti-PCMT1 antibody produced in rabbit is suitable for use in proteome-wide epitope mapping covering all human proteins for on- and off-target binding analysis.
Anti-PCMT1 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

PCMT1 (protein-L-isoaspartate (D-aspartate) O-methyltransferase) protein repairs modified proteins by catalyzing the methyl esterification of L-isoaspartyl and D-aspartyl residues in peptides and proteins that are formed from spontaneous decomposition of normal L-aspartyl and L-asparaginyl residues. It negatively regulates p53, a tumour suppressor protein, and in turn downregulates p53-mediated transcription of target genes. It may serve as a therapeutic target for cancers. It protects neural cells from Bax-induced apoptosis and defects in this gene may cause spina bifida in infants. It activates integrin αv and mediates cell adhesion in various cancer cell lines. The protein negatively regulates β−amyloid peptide formation and plays a protective role in Alzheimer′s disease pathogenesis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST79907

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Feng Liang et al.
Scientific reports, 7(1), 9201-9201 (2017-08-25)
Neuronal apoptosis chiefly contributes to the cell loss following traumatic brain injury (TBI). CGP3466B is a compound related to the anti-Parkinsonism drug R-(-)-deprenyl. Previous studies have illuminated anti-apoptosis effects of CGP3466B in different cell lines, but the underlying mechanisms have
Björn Forsström et al.
Molecular & cellular proteomics : MCP, 13(6), 1585-1597 (2014-04-08)
Antibodies are of importance for the field of proteomics, both as reagents for imaging cells, tissues, and organs and as capturing agents for affinity enrichment in mass-spectrometry-based techniques. It is important to gain basic insights regarding the binding sites (epitopes)
Jae-Cheol Lee et al.
Nature communications, 3, 927-927 (2012-06-28)
Protein methylation plays important roles in most, if not all, cellular processes. Lysine and arginine methyltransferases are known to regulate the function of histones and non-histone proteins through the methylation of specific sites. However, the role of the carboxyl-methyltransferase protein
Ana M Wägner et al.
The review of diabetic studies : RDS, 5(4), 225-231 (2009-03-19)
Posttranslational protein modifications have been implicated in the development of autoimmunity. Protein L-isoaspartate (D-aspartate) O-methyltransferase (PIMT) repairs modified proteins and is encoded by PCMT1, located in a region linked to type 1 diabetes (T1D), namely IDDM5. To evaluate the association
Jiyeon Ryu et al.
Acta pharmacologica Sinica, 32(9), 1165-1172 (2011-08-16)
Protein L-isoaspartyl O-methyltransferase (PIMT) regulates cell adhesion in various cancer cell lines through activation of integrin αv and the PI3K pathway. The epithelial mesenchymal transition (EMT) enables epithelial cells to acquire the characteristics of mesenchymal cells, and to allow them

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service