Skip to Content
Merck
All Photos(10)

Key Documents

HPA001909

Sigma-Aldrich

Anti-LAMC1 antibody produced in rabbit

enhanced validation

Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Laminin B2 chain antibody produced in rabbit, Anti-Laminin subunit γ-1 precursor antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human, mouse

enhanced validation

independent
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

STKAEAERTFAEVTDLDNEVNNMLKQLQEAEKELKRKQDDADQDMMMAGMASQAAQEAEINARKAKNSVTSLLSIINDLLEQLGQLDTVDLNKLNEIEGTLNKAKDEMKVS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LAMC1(3915)

Looking for similar products? Visit Product Comparison Guide

General description

Laminin is a large 900kDa complex mosaic glycoprotein synthesized by a range of cells and is deposited in basement membranes. LAMC1 (laminin subunit γ-1) is a component of laminin. It contains a cysteine-rich repeat and a globular region. LAMC1 is associated with various biological functions such as cell attachment, mitogenesis, migration, and cell differentiation. It is upregulated in high-grade serous carcinomas (HGSCs).

Immunogen

Laminin subunit γ-1 precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-LAMC1 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86216

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

You-Quan Wang et al.
Breast cancer research and treatment, 188(1), 1-14 (2021-03-31)
Breast cancer (BC) is the most prevalent cancer in women with an estimated incidence of 10% and the leading cause of mortality due to its heterogenous property and high metastasis rate. Development of novel therapy is very necessary and requires
Kala Visvanathan et al.
Cancer prevention research (Philadelphia, Pa.), 11(11), 697-706 (2018-09-21)
The prognosis of women diagnosed with invasive high-grade serous ovarian carcinoma (HGSC) is poor. More information about serous tubal intraepithelial carcinoma (STIC) and serous tubal intraepithelial lesions (STIL), putative precursor lesions of HGSC, could inform prevention efforts. We conducted a
Lin-Yu Chen et al.
The Journal of pathology, 248(3), 363-376 (2019-03-19)
Ten-eleven translocation methylcytosine dioxygenase-1, TET1, takes part in active DNA demethylation. However, our understanding of DNA demethylation in cancer biology and its clinical significance remain limited. This study showed that TET1 expression correlated with poor survival in advanced-stage epithelial ovarian
K Beck et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 4(2), 148-160 (1990-02-01)
Laminin is a large (900 kDa) mosaic protein composed of many distinct domains with different structures and functions. Globular and rodlike domains are arranged in an extended four-armed, cruciform shape that is well suited for mediating between distant sites on
Jennifer X Ji et al.
The Journal of pathology, 258(4), 325-338 (2022-08-30)
Clear cell ovarian carcinoma (CCOC) is the second most common subtype of epithelial ovarian carcinoma. Late-stage CCOC is not responsive to gold-standard chemotherapy and results in suboptimal outcomes for patients. In-depth molecular insight is urgently needed to stratify the disease

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service