Skip to Content
Merck
All Photos(1)

Key Documents

AB3553

Sigma-Aldrich

Anti-Thiazide-Sensitive NaCl Cotransporter Antibody

Chemicon®, from rabbit

Synonym(s):

Anti-NCC, Anti-NCCT, Anti-TSC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rabbit, mouse, rat, human

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SLC12A3(6559)

Specificity

Recognizes the renal thiazide-sensitive NaCl cotransporter (NCC). The antibody stains the apical plasma membrane and subapical membrane vesicles in renal distal convoluted tubule cells.

Immunogen

Synthetic peptide from the N-terminal of human/rat/mouse NCC.

Application

Immunoblotting: 1:2,000 using rat kidney extract. The antibody reacts with the 160-190 kDa protein (the molecular weight varies depending on conditions for protein extraction/electrophoresis). The suggested dilution and blocking buffer is 150 mM NaCl, 10 mM Tris, pH 7.4 containing 5% low fat milk and 0.04% Tween. Suggested transfer membrane is PVDF or Nylon. Suggested gel percentage is 7%. Overnight incubation with the antibody at 2-8°C is recommended.

Immunohistochemistry: 1:500-1:2,000 on rat or mouse kidney tissue. Suggested fixative is 3% paraformaldehyde for 5 minutes by vascular perfusion. Suggested blocking buffer is PBS containing 10% normal goat serum. Suggested dilution buffer is PBS containing 1% BSA. Overnight incubation with the antibody at 2-8°C is recommended.Optimal working dilutions must be determined by the end user.
Research Category
Neuroscience
Research Sub Category
Ion Channels & Transporters
This Anti-Thiazide-Sensitive NaCl Cotransporter Antibody is validated for use in IH, WB for the detection of Thiazide-Sensitive NaCl Cotransporter.

Target description

160-190 kDa

Linkage

Replaces: AB3564P

Physical form

Affinity Purified immunoglobulin in PBS containing no preservatives.
ImmunoAffinity Purified

Storage and Stability

Maintain for 1 year at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
POSITIVE CONTROL: Rat kidney.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Renal tubule angiotensin II type 1 receptor-associated protein promotes natriuresis and inhibits salt-sensitive blood pressure elevation.
Wakui, H; Uneda, K; Tamura, K; Ohsawa, M; Azushima, K; Kobayashi, R; Ohki, K; Dejima et al.
Journal of the American Heart Association null
Smail Messaoudi et al.
Nature communications, 6, 8835-8835 (2015-12-01)
Despite its high prevalence and economic burden, the aetiology of human hypertension remains incompletely understood. Here we identify the transcription factor GATA5, as a new regulator of blood pressure (BP). GATA5 is expressed in microvascular endothelial cells and its genetic
Genomic-derived markers for early detection of calcineurin inhibitor immunosuppressant-mediated nephrotoxicity.
Cui, Y; Huang, Q; Auman, JT; Knight, B; Jin, X; Blanchard, KT; Chou, J; Jayadev, S; Paules, RS
Toxicological Sciences null
Omar A Z Tutakhel et al.
PloS one, 12(4), e0176220-e0176220 (2017-04-22)
Animal studies have shown that the calcineurin inhibitors (CNIs) cyclosporine and tacrolimus can activate the thiazide-sensitive NaCl cotransporter (NCC). A common side effect of CNIs is hypertension. Renal salt transporters such as NCC are excreted in urinary extracellular vesicles (uEVs)
Jessica R Ivy et al.
Hypertension (Dallas, Tex. : 1979), 67(5), 1029-1037 (2016-03-10)
Blood pressure (BP) normally dips during sleep, and nondipping increases cardiovascular risk. Hydrochlorothiazide restores the dipping BP profile in nondipping patients, suggesting that the NaCl cotransporter, NCC, is an important determinant of daily BP variation. NCC activity in cells is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service