Skip to Content
Merck
All Photos(2)

Key Documents

H3159

Sigma-Aldrich

Anti-Histone Deacetylase 2 (HDAC2) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-HD2, Anti-KDAC2, Anti-RPD3, Anti-YAF1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 55 kDa

species reactivity

mouse, human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:500 using protease-digested, human lymph node sections
immunoprecipitation (IP): 50 μg using whole lysate of NIH3T3 cells
microarray: suitable
western blot: 1:20,000 using nuclear extract from human HeLa cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HDAC2(3066)
mouse ... Hdac2(15182)

General description

Histone Deacetylase (HDACs) have been described as six or seven different groups in mammals. HDAC1, HDAC2 and HDAC3 are like yeast Rpd3 protein, while HDAC4, HDAC5 and HDAC6 are like yeast Hda1 (Histone Deacetylase 1) protein.

Specificity

Anti-Histone Deacetylase 2 specifically recognizes histone deacetylase 2 by immunoblotting and immunoprecipitation (55 kDa). An additional band of lower molecular weight may be detected in some cell line extracts by immunoblotting. Staining of HDAC2 by immunoblotting is specifically inhibited with the immunizing peptide. The product is also useful for the detection of HDAC2 by immunohistochemistry. The epitope(s) recognized by the antibody is resistant to routine formalin-fixation and paraffin embedding. The antibody reacts with HDAC2 of human, rat and mouse origin.

Immunogen

synthetic peptide (C-SGEKTDTKGTKSEQLSNP) corresponding to the C-terminus region of histone deacetylase 2 of human origin (amino acids residues 471-488 with N-ternimal added cysteine) conjugated to maleimide-activated KLH. This sequence is highly conserved in mouse and chicken (2 amino acids and 1 amino acid substitutions, respectively).

Application

Anti-Histone Deacetylase 2 (HDAC2) antibody has been used:
  • in immunoblotting and dot blot
  • in immunoprecipitation
  • in immunohistochemistry
  • in enzyme linked immunosorbent assay (ELISA)
  • in western blotting
  • in chromatin immunoprecipitation(ChiP)

Biochem/physiol Actions

Histone acetylation is a dynamic process whose levels are determined by the net activities of Histone acetyl transferase (HATs) and the competing enzymes histone deacetylases (HDACs). Both activities are associated with the nuclear matrix. Histone deacetylases activities were often, but not always, associated with transcriptional repression and nucleosome condensations. HDAC1, HDAC2 and several other HDACs are the catalytic subunits of different multiprotein regulatory complexes.

Physical form

Solution in 0.01 M phosphate bufffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development of an ELISA-based HDAC activity assay for characterization of isoform-selective inhibitors
Padige G, et al.
Journal of Biomolecular Screening, 20(10), 1277-1285 (2015)
The primary microRNA-208b interacts with Polycomb-group protein, Ezh2, to regulate gene expression in the heart
Mathiyalagan P, et al.
Nucleic Acids Research, 42(2), 790-803 (2013)
Anticancer activities of histone deacetylase inhibitors
Bolden J E, et al.
Nature Reviews Drug Discovery, 5(9), 769-769 (2006)
DNA methyltransferase 3b preferentially associates with condensed chromatin
Kashiwagi K, et al.
Nucleic Acids Research, 39(3), 874-888 (2010)
SAHA decreases HDAC 2 and 4 levels in vivo and improves molecular phenotypes in the R6/2 mouse model of Huntington's disease
Mielcarek M, et al.
PLoS ONE, 6(11), e27746-e27746 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service