Skip to Content
Merck
All Photos(1)

Key Documents

G9152

Sigma-Aldrich

Anti-Galactocerebroside antibody produced in rabbit

whole antiserum

Synonym(s):

Galactocerebroside Antibody, Galactocerebroside Antibody - Anti-Galactocerebroside antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
DB
clone:
polyclonal
species reactivity:
human
citations:
15
technique(s):
dot blot: 1:50 using galactocerebroside I and II

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

whole antiserum

antibody product type

primary antibodies

clone

polyclonal

contains

15 mM sodium azide

species reactivity

human

technique(s)

dot blot: 1:50 using galactocerebroside I and II

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GALC(2581)

General description

Galactocerebroside is the major galactosphingolipid of myelin. It is expressed in the CNS in cell membranes of oligodendrocytes. It is also expressed in Schwann cells in peripheral nerves.
This product is a pooled delipidized antiserum produced in rabbits by repeated injections of a mixture of galactocerebrosides I and II conjugated to keyhole limpet hemocyanin.

Specificity

The antibody localizes oligodendroglia in cultured cells. It may be used for studying changes in the organization of oligodendroglial membrane sheets in culture. Specificity of the product is determined by dot blotting using galactocerebroside as the antigen.

Immunogen

Galactocerebrosides (GalC) I and II conjugated to KLH

Application

Anti-Galactocerebroside antibody produced in rabbit has been used:
  • for dot-blot immunoassay and western blot analysis of mouse brain homogenates
  • as a primary antibody at a working dilution of 1:50 for immunostaining of muscle tissue of Rana pipiens frogs
  • as a primary antibody at a working dilution of 1:500 in immunofluorescence and immunohistochemistry using primary human astroglia

Biochem/physiol Actions

Galactocerebroside is the major galactosphingolipid of myelin. It is expressed in the CNS in cell membranes of oligodendrocytes. It is also expressed in Schwann cells in peripheral nerves. Galactocerebroside is found in serum and cerebral spinal fluid of patients with demyelinating diseases like multiple sclerosis.
Galactocerebroside was shown to play an important role in the processes of normal development and myelination and is known to be an early marker of oligodendrocyte development.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D M Kokkinakis et al.
Neuro-oncology, 3(2), 99-112 (2001-04-12)
Glial tumors may originate from the malignant transformation of multipotent glial progenitor cells, but tools to study malignant transformation leading to gliomas are limited by the lack of biological systems that represent early stages of this disease in adult animals.
C A Dyer et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 8(11), 4307-4318 (1988-11-01)
Antibodies to galactocerebroside (GalC) cause major changes in the organization of the membrane sheets elaborated by murine oligodendroglia in culture. Exposure of oligodendroglia to rabbit anti-GalC IgG for 15 min followed by fluoresceinated second antibodies results in patches of surface
J W Prineas et al.
Laboratory investigation; a journal of technical methods and pathology, 61(5), 489-503 (1989-11-01)
Fresh lesions in the brain and spinal cord of patients with multiple sclerosis who died shortly after the onset of symptoms were examined immunocytochemically for myelin and oligodendrocyte antigens that are known to be sequentially expressed during normal development. Cells
Lead alters the developmental profile of the galactolipid metabolic enzymes in cultured oligodendrocyte lineage cells
Deng W and Poretz RD
Neurotoxicology, 22(4), 429-437 (2001)
J Georgiou et al.
Glia, 27(2), 101-109 (1999-07-27)
Perisynaptic Schwann cells (PSCs) envelop axonal terminals and are physiologically distinct from the nearby myelinating Schwann cells (MSCs), which surround the same innervating motor axons. PSCs have special functions at the neuromuscular synapse, where they detect and can modulate neurotransmitter

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service