Skip to Content
Merck
All Photos(1)

Documents

G6772

Sigma-Aldrich

Geranyl pyrophosphate ammonium salt

≥95% (TLC), liquid

Synonym(s):

trans-3,7-Dimethyl-2,6-octadienyl pyrophosphate, GPP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H20O7P2 · 3NH3
CAS Number:
Molecular Weight:
365.30
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

product name

Geranyl pyrophosphate ammonium salt, 1 mg/mL in methanol (:aqueous 10 mM NH4OH (7:3)), ≥95% (TLC)

Quality Level

Assay

≥95% (TLC)

form

liquid

packaging

vial of 200 μg

concentration

1 mg/mL in methanol (:aqueous 10 mM NH4OH (7:3))

storage temp.

−20°C

SMILES string

C\C(C)=C\CC\C(C)=C\COP(O)(=O)OP(O)(O)=O

InChI

1S/C10H20O7P2/c1-9(2)5-4-6-10(3)7-8-16-19(14,15)17-18(11,12)13/h5,7H,4,6,8H2,1-3H3,(H,14,15)(H2,11,12,13)/b10-7+

InChI key

GVVPGTZRZFNKDS-JXMROGBWSA-N

Related Categories

Biochem/physiol Actions

Intermediate in terpene biosynthesis

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

68.0 °F - closed cup

Flash Point(C)

20 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wenping Yuan et al.
Science advances, 5(8), eaax1396-eaax1396 (2019-08-28)
Atmospheric vapor pressure deficit (VPD) is a critical variable in determining plant photosynthesis. Synthesis of four global climate datasets reveals a sharp increase of VPD after the late 1990s. In response, the vegetation greening trend indicated by a satellite-derived vegetation
Iryna Janssen et al.
International journal of medical microbiology : IJMM, 306(8), 652-656 (2016-10-04)
Since data about Clostridium difficile infection in sub-Saharan Africa are scarce, we determined its epidemiology and risk factors in a cross-sectional study in Eikwe, a rural community in Ghana. We tested stool samples from 176 hospitalized patients with diarrhoea and
Jacob Schewe et al.
Nature communications, 10(1), 1005-1005 (2019-03-03)
Global impact models represent process-level understanding of how natural and human systems may be affected by climate change. Their projections are used in integrated assessments of climate change. Here we test, for the first time, systematically across many important systems
Sophie Twelves et al.
The Journal of allergy and clinical immunology, 143(3), 1021-1026 (2018-07-24)
The term pustular psoriasis indicates a group of severe skin disorders characterized by eruptions of neutrophil-filled pustules. The disease, which often manifests with concurrent psoriasis vulgaris, can have an acute systemic (generalized pustular psoriasis [GPP]) or chronic localized (palmoplantar pustulosis
M Fernández-Martínez et al.
Scientific reports, 7(1), 9632-9632 (2017-08-31)
Concentrations of atmospheric carbon dioxide (CO

Articles

Cholesterol biosynthesis starts in the hepatic endoplasmic reticulum with acetyl-CoA, yielding 3-hydroxy-3-methylglutaryl-CoA via HMG-CoA synthase.

Cholesterol biosynthesis starts in the hepatic endoplasmic reticulum with acetyl-CoA, yielding 3-hydroxy-3-methylglutaryl-CoA via HMG-CoA synthase.

Cholesterol biosynthesis starts in the hepatic endoplasmic reticulum with acetyl-CoA, yielding 3-hydroxy-3-methylglutaryl-CoA via HMG-CoA synthase.

Cholesterol biosynthesis starts in the hepatic endoplasmic reticulum with acetyl-CoA, yielding 3-hydroxy-3-methylglutaryl-CoA via HMG-CoA synthase.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service