Skip to Content
Merck

M9410

Nunc-Immuno MicroWell 96 well solid plates

96 well MicroWell MaxiSorp flat bottom plate, pinchbar design

Synonym(s):

96 well plates, cell culture plates, tissue culture plates, Nunc 442404, ELISA plates, cell culture plate, microwell plates, multiwell plates

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.22
UNSPSC Code:
41121800
Material:
flat bottom clear polystyrene wells, polystyrene
Size:
96 wells
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

flat bottom clear polystyrene wells, polystyrene

sterility

non-sterile

feature

lid: no, skirt

packaging

case of 60 ea

manufacturer/tradename

Nunc 442404

plate L × W

128 mm × 86 mm

size

96 wells

surface area

2.7 cm2

total capacity

400 μL

well maximum volume

400 μm

working volume

350 μL

suitability

suitable for (quantitative/qualitative solid phase immuno assays (e.g. ELISA) and binding assays)

binding type

(MaxiSorp surface), high binding surface

General description

Nunc-Immuno 96 MicroWell solid plates are intended for quantitative and qualitative solid phase immuno assays (i.e. ELISA) and binding assays.


Surface Treaments:


  • PolySorp® surface treament, high affinity to molecules of a hydrophobic nature
  • MediSorp surface treament, surface chemistry between PolySorp and MaxiSorp, provides low background signal with samples containing serum
  • MaxiSorp surface treament, high affinity to molecules with mixed hydrophilic/hydrophobic domains
  • MultiSorp surface treament, high affinity to hydrophilic molecules

Application

Nunc-Immuno 96 MicroWell solid plates have been used for quantitative and qualitative solid phase immuno assays, such as ELISA.

Features and Benefits

  • Raised rims on the wells
  • Certified reproducibility of binding
  • Alphanumeric orientation system
  • Fit standard equipment
  • Barcodes options available

Legal Information

MaxiSorp is a trademark of Thermo Fisher Scientific or its subsidiaries
MediSorp is a trademark of Thermo Fisher Scientific or its subsidiaries
MicroWell is a trademark of Thermo Fisher Scientific or its subsidiaries
MultiSorp is a trademark of Thermo Fisher Scientific or its subsidiaries
Nunc-Immuno is a trademark of Thermo Fisher Scientific or its subsidiaries
PolySorp is a registered trademark of Thermo Fisher Scientific or its subsidiaries


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library