Skip to Content
Merck
All Photos(1)

Key Documents

F6771

Sigma-Aldrich

Fumagillin

from Aspergillus sp. (Aspergillus fumigatus), ≥90% (HPLC), powder, MetAP2 inhibitor

Synonym(s):

Fugillin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H34O7
CAS Number:
Molecular Weight:
458.54
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Product Name

Fumagillin from Aspergillus fumigatus, ≥90%, powder

biological source

Aspergillus sp. (Aspergillus fumigatus)

Quality Level

Assay

≥90%

form

powder

color

white

mp

197-198  °C

solubility

ethanol: 1 mg/mL

antibiotic activity spectrum

fungi

Mode of action

DNA synthesis | interferes

storage temp.

−20°C

SMILES string

COC1C(CCC2(CO2)C1C3(C)OC3C\C=C(\C)C)OC(=O)\C=C\C=C\C=C\C=C\C(O)=O

InChI

1S/C26H34O7/c1-18(2)13-14-20-25(3,33-20)24-23(30-4)19(15-16-26(24)17-31-26)32-22(29)12-10-8-6-5-7-9-11-21(27)28/h5-13,19-20,23-24H,14-17H2,1-4H3,(H,27,28)/b7-5+,8-6+,11-9+,12-10+

InChI key

NGGMYCMLYOUNGM-HCNIIHBUSA-N

General description

Fumagillin is a metabolic product of fungi. It is originally isolated from Aspergillus fumigatus.

Application

Fumagillin from Aspergillus fumigatus has been used:
  • in chick chorioallantoic membrane (CAM) assay to study the changes in protein expression in response to anti-angiogenic agent
  • to study its effects on the mortality of caged Nosema ceranae-infected honey bees- write in this way
  • as a positive control in in vitro and ex vivo studies

Biochem/physiol Actions

Fumagillin is known to be effective against microsporidian fungal infections.
Methionine aminopeptidase-2 (MetAP-2) inhibitor; inhibits endothelial cell proliferation and angiogenesis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Fumagillin: an overview of recent scientific advances and their significance for apiculture
van den Heever JP, et al.
Journal of Agricultural and Food Chemistry, 62(13), 2728-2737 (2014)
The effect of dicyclohexylamine and fumagillin on Nosema ceranae-infected honey bee (Apis mellifera) mortality in cage trial assays
Van den Heever JP, et al.
Apidologie, 47(5), 663-670 (2016)
A novel technique for quantifying changes in vascular density, endothelial cell proliferation and protein expression in response to modulators of angiogenesis using the chick chorioallantoic membrane (CAM) assay
Miller WJ, et al.
Journal of Translational Medicine, 2(1), 4-4 (2004)
Catherine Z Chen et al.
Antimicrobial agents and chemotherapy, 55(2), 667-675 (2010-11-17)
The human pathogen Giardia lamblia is an anaerobic protozoan parasite that causes giardiasis, one of the most common diarrheal diseases worldwide. Although several drugs are available for the treatment of giardiasis, drug resistance has been reported and is likely to
Hsiao-Ching Lin et al.
Journal of the American Chemical Society, 135(12), 4616-4619 (2013-03-16)
Fumagillin 1 is a meroterpenoid from Aspergillus fumigatus that is known for its anti-angiogenic activity by binding to human methionine aminopeptidase 2. The genetic and molecular basis for biosynthesis of 1 had been an enigma despite the availability of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service