Skip to Content
Merck
All Photos(1)

Key Documents

F0429

Sigma-Aldrich

Flunixin meglumine

≥98% (HPLC)

Synonym(s):

2-[[2-Methyl-3-(trifluoromethyl)phenyl]amino]-3-pyridinecarboxylic acid meglumine salt, Banamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H11F3N2O2·C7H17NO5
CAS Number:
Molecular Weight:
491.46
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

mp

136.6-137.4 °C

solubility

H2O: freely soluble

storage temp.

room temp

SMILES string

CNC[C@H](O)[C@@H](O)[C@H](O)[C@H](O)CO.Cc1c(Nc2ncccc2C(O)=O)cccc1C(F)(F)F

InChI

1S/C14H11F3N2O2.C7H17NO5/c1-8-10(14(15,16)17)5-2-6-11(8)19-12-9(13(20)21)4-3-7-18-12;1-8-2-4(10)6(12)7(13)5(11)3-9/h2-7H,1H3,(H,18,19)(H,20,21);4-13H,2-3H2,1H3/t;4-,5+,6+,7+/m.0/s1

InChI key

MGCCHNLNRBULBU-WZTVWXICSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Flunixin meglumine (IC50 = 1 nM) can be used as a drug for animals for the management of intestinal ischaemia, colic, and endotoxemia in horses.
Flunixin meglumine has been used as a nonsteroidal anti-inflammatory drug standard in electrospray ionization mass spectrometry (LC-ESI/MS).

Biochem/physiol Actions

COX1, COX2 and PLA2 inhibitor non-narcotic analgesic, anti-pyretic, anti-inflammatory.
Flunixin meglumine is a nonsteroidal anti-inflammatory drug, which blocks prostaglandin synthesis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

A Review on Flunixin Meglumine
Faiza H, et al.
International journal of medicine and medical research, 1(2), 136-138 (2016)
Analysis of nine NSAIDs in ungulate tissues available to critically endangered vultures in India
Taggart MA, et al.
Environmental Science & Technology, 43(12), 4561-4566 (2009)
J Cuervo-Arango et al.
Theriogenology, 77(3), 514-524 (2011-10-01)
The development of hemorrhagic anovulatory follicles (HAF) involves luteinization and hemorrhage of the follicle. This is observed on ultrasound as an increase in the echogenicity of the granulosa layer and formation of echoic particles in the antrum. The inhibition of
G Pugliesi et al.
Theriogenology, 78(2), 443-454 (2012-05-15)
The beginning of postluteolysis (progesterone, <1 ng mL(-1)) in heifers was targeted by using 8 h after ultrasonic detection of a 25% decrease in CL area (cm2) and was designated Hour 0. Flunixin meglumine (FM; n=10) to inhibit PGF2α secretion
C Donalisio et al.
Journal of veterinary pharmacology and therapeutics, 36(2), 130-139 (2012-06-26)
In this study, ex vivo assays were carried out in dairy cows to evaluate the anti-inflammatory effects of two nonsteroidal anti-inflammatory drugs: ketoprofen (KETO) and flunixin meglumine (FM). Twelve healthy Holstein dairy cattle were randomly allocated to two groups (n=6):

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service