Skip to Content
Merck
All Photos(1)

Documents

D7770

Supelco

Diminazene aceturate

analytical standard

Synonym(s):

Berenil

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H15N7 · 2C4H7NO3
CAS Number:
Molecular Weight:
515.52
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

form

powder

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

SMILES string

CC(=O)NCC(O)=O.CC(=O)NCC(O)=O.NC(=N)c1ccc(N\N=N\c2ccc(cc2)C(N)=N)cc1

InChI

1S/C14H15N7.2C4H7NO3/c15-13(16)9-1-5-11(6-2-9)19-21-20-12-7-3-10(4-8-12)14(17)18;2*1-3(6)5-2-4(7)8/h1-8H,(H3,15,16)(H3,17,18)(H,19,20);2*2H2,1H3,(H,5,6)(H,7,8)

InChI key

OKQSSSVVBOUMNZ-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Gysin et al.
International journal for parasitology. Drugs and drug resistance, 8(2), 159-164 (2018-03-28)
Trypanosoma congolense is a protozoan parasite that is transmitted by tsetse flies, causing African Animal Trypanosomiasis, also known as Nagana, in sub-Saharan Africa. Nagana is a fatal disease of livestock that causes severe economic losses. Two drugs are available, diminazene
Jia Yu Peppermint Lee et al.
British journal of pharmacology, 175(12), 2204-2218 (2017-11-15)
Acid-sensing ion channels (ASICs) are primary acid sensors in mammals, with the ASIC1b and ASIC3 subtypes being involved in peripheral nociception. The antiprotozoal drug diminazene is a moderately potent ASIC inhibitor, but its analgesic activity has not been assessed. We
Thiago Souza Azeredo Bastos et al.
Revista brasileira de parasitologia veterinaria = Brazilian journal of veterinary parasitology : Orgao Oficial do Colegio Brasileiro de Parasitologia Veterinaria, 26(3), 366-371 (2017-07-06)
Trypanosomiasis caused by Trypanosoma vivax has increased the reports in Brazil in the last decade. An outbreak is herein first reported in the state of Goiás, from May 2016 to January 2017. The outbreak start occurred in the city of
Raqueli T França et al.
Veterinary clinical pathology, 42(1), 31-39 (2013-03-06)
Rangelia vitalii is a tick-transmitted piroplasm that causes both hemolytic and hemorrhagic disease in dogs in Brazil. The aim of this study was to evaluate the response of the bone marrow in dogs experimentally infected with R vitalii during the
Aboagye Kwarteng Dofuor et al.
Biomolecules, 10(12) (2020-12-17)
In the absence of vaccines, there is a need for alternative sources of effective chemotherapy for African trypanosomiasis (AT). The increasing rate of resistance and toxicity of commercially available antitrypanosomal drugs also necessitates an investigation into the mode of action

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service