Skip to Content
Merck
All Photos(1)

Key Documents

E6877

Sigma-Aldrich

[Ala1,3,11,15]-Endothelin 1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C109H163N25O32S
CAS Number:
Molecular Weight:
2367.67
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32

storage temp.

−20°C

Quality Level

SMILES string

CC[C@H](C)[C@H](NC(=O)[C@H](CC(O)=O)NC(=O)[C@H](CC(C)C)NC(=O)[C@H](Cc1c[nH]cn1)NC(=O)[C@H](C)NC(=O)[C@H](Cc2ccccc2)NC(=O)[C@H](Cc3ccc(O)cc3)NC(=O)[C@@H](NC(=O)[C@H](C)NC(=O)[C@H](CCC(O)=O)NC(=O)[C@H](CCCCN)NC(=O)[C@H](CC(O)=O)NC(=O)[C@H](CCSC)NC(=O)[C@H](CC(C)C)NC(=O)[C@H](CO)NC(=O)[C@H](CO)NC(=O)[C@H](C)NC(=O)[C@H](CO)NC(=O)[C@H](C)N)C(C)C)C(=O)N[C@@H]([C@@H](C)CC)C(=O)N[C@@H](Cc4c[nH]c5ccccc45)C(O)=O

InChI

1S/C109H163N25O32S/c1-16-56(9)87(107(163)128-79(109(165)166)43-64-47-113-68-28-22-21-27-67(64)68)134-108(164)88(57(10)17-2)133-102(158)78(46-85(143)144)126-98(154)73(40-54(5)6)122-100(156)76(44-65-48-112-52-114-65)121-90(146)59(12)116-96(152)74(41-62-25-19-18-20-26-62)124-99(155)75(42-63-30-32-66(138)33-31-63)127-106(162)86(55(7)8)132-92(148)61(14)115-93(149)70(34-35-83(139)140)119-94(150)69(29-23-24-37-110)118-101(157)77(45-84(141)142)125-95(151)71(36-38-167-15)120-97(153)72(39-53(3)4)123-104(160)82(51-137)131-105(161)81(50-136)130-91(147)60(13)117-103(159)80(49-135)129-89(145)58(11)111/h18-22,25-28,30-33,47-48,52-61,69-82,86-88,113,135-138H,16-17,23-24,29,34-46,49-51,110-111H2,1-15H3,(H,112,114)(H,115,149)(H,116,152)(H,117,159)(H,118,157)(H,119,150)(H,120,153)(H,121,146)(H,122,156)(H,123,160)(H,124,155)(H,125,151)(H,126,154)(H,127,162)(H,128,163)(H,129,145)(H,130,147)(H,131,161)(H,132,148)(H,133,158)(H,134,164)(H,139,140)(H,141,142)(H,143,144)(H,165,166)/t56-,57-,58-,59-,60-,61-,69-,70-,71-,72-,73-,74-,75-,76-,77-,78-,79-,80-,81-,82-,86-,87-,88-/m0/s1

InChI key

NRTWVLDGYXCWHD-IGKRDWOESA-N

Gene Information

human ... EDNRB(1910)
mouse ... EDNRB(13618)
rat ... EDNRB(50672)

Amino Acid Sequence

Ala-Ser-Ala-Ser-Ser-Leu-Met-Asp-Lys-Glu-Ala-Val-Tyr-Phe-Ala-His-Leu-Asp-Ile-Ile-Trp

General description

[Ala1,3,11,15]-Endothelin 1 is a linear endothelin (ET)-1 analogue, and acts as an ETB receptor agonist. ET-1 is a peptide that acts as both, a vasodilator and a vasoconstrictor. It is a ligand for ET receptor, which is of two types, ETA and ETB. ETB receptor is found in the vascular endothelial cells of cardiovascular tissues.

Application

[Ala1,3,11,15]-Endothelin 1 may be used to assess the effects of endothelin-1 and [Ala1,3,11,15]-endothelin 1 produced by ETA and ETB receptors, in the presence of BQ-123, in anaesthetized rat.

Biochem/physiol Actions

Selective agonist for ETB endothelin receptors.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P S Widdowson et al.
British journal of pharmacology, 118(8), 2126-2130 (1996-08-01)
1. We performed radioligand binding experiments on rat cerebellar homogenates using [125I]-endothelin-1 ¿[1251]-ET-1¿ and [125I]-BQ3020 to examine the pharmacology of endothelin receptors in rat brain. Saturation experiments demonstrated a single population of binding sites with high affinity for both radioligands
J Green
International journal of peptide and protein research, 41(5), 492-498 (1993-05-01)
[Ala1,3,11,15]-Endothelin-1, a linear analogue of endothelin-1 in which alanines replace the cystine residues, has been prepared by solid-phase synthesis in approximately 17% yield. Fmoc amino acids were coupled using an economical modified double-coupling (symmetrical anhydride followed by O-benzotriazolyl ester) procedure
S Topouzis et al.
British journal of pharmacology, 98(2), 669-677 (1989-10-01)
1. The aim of the present study was to examine the contractile responses evoked by the recently characterized vasoactive peptide endothelin-1 (ET-1) and by two of its structural analogues, [Ala3,11]ET-1 and [Ala1,15]ET-1 in endothelium-denuded rat isolated aorta, and also to
A D Wren et al.
Biochemical and biophysical research communications, 196(1), 369-375 (1993-10-15)
An in vitro model of endothelial cell injury was used to investigate the role of endothelins and related peptides in endothelial repair. Endothelin-3 (10-100 nM) enhanced wound repair over an 18 h period by promoting proliferation, an effect not inhibited
M D Randall et al.
British journal of pharmacology, 98(2), 685-699 (1989-10-01)
1. The effects of endothelin-1 and of three analogues in which alanyl residues had been substituted in place of cysteinyl residues were studied in the rat, isolated, Krebs-Henseleit-perfused mesenteric bed and the in situ, blood-perfused, mesenteric and hindquarters circulations of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service