Skip to Content
Merck

Skip To

E7764

Endothelin 1

≥97% (HPLC), powder

Synonym(s):

ET-1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C109H159N25O32S5
CAS Number:
Molecular Weight:
2491.90
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Form:
powder
Assay:
≥97% (HPLC)
Biological source:
human
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human

Quality Segment

assay

≥97% (HPLC)

form

powder

technique(s)

ELISA: suitable

color

white

solubility

1% acetic acid: >1 mg/mL, H2O: >1 mg/mL

UniProt accession no.

storage temp.

−20°C

SMILES string

[H]N[C@H]1CSSC[C@H](NC(=O)[C@H](Cc2ccccc2)NC(=O)[C@H](Cc3ccc(O)cc3)NC(=O)[C@@H](NC(=O)[C@@H]4CSSC[C@H](NC(=O)[C@H](CO)NC1=O)C(=O)N[C@@H](CO)C(=O)N[C@@H](CO)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCSC)C(=O)N[C@@H](CC(O)=O)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CCC(O)=O)C(=O)N4)C(C)C)C(=O)N[C@@H](Cc5cnc[nH]5)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CC(O)=O)C(=O)N[C@@H]([C@@H](C)CC)C(=O)N[C@@H]([C@@H](C)CC)C(=O)N[C@@H](Cc6c[nH]c7ccccc67)C(O)=O

InChI

1S/C109H159N25O32S5/c1-12-56(9)87(107(163)125-76(109(165)166)39-60-43-113-65-24-18-17-23-63(60)65)134-108(164)88(57(10)13-2)133-99(155)75(42-85(143)144)123-94(150)70(36-54(5)6)118-97(153)73(40-61-44-112-52-114-61)121-103(159)80-49-169-168-48-64(111)89(145)126-77(45-135)102(158)131-81-50-170-171-51-82(105(161)132-86(55(7)8)106(162)124-72(38-59-26-28-62(138)29-27-59)95(151)120-71(96(152)130-80)37-58-21-15-14-16-22-58)129-91(147)67(30-31-83(139)140)116-90(146)66(25-19-20-33-110)115-98(154)74(41-84(141)142)122-92(148)68(32-34-167-11)117-93(149)69(35-53(3)4)119-100(156)78(46-136)127-101(157)79(47-137)128-104(81)160/h14-18,21-24,26-29,43-44,52-57,64,66-82,86-88,113,135-138H,12-13,19-20,25,30-42,45-51,110-111H2,1-11H3,(H,112,114)(H,115,154)(H,116,146)(H,117,149)(H,118,153)(H,119,156)(H,120,151)(H,121,159)(H,122,148)(H,123,150)(H,124,162)(H,125,163)(H,126,145)(H,127,157)(H,128,160)(H,129,147)(H,130,152)(H,131,158)(H,132,161)(H,133,155)(H,134,164)(H,139,140)(H,141,142)(H,143,144)(H,165,166)/t56-,57-,64-,66-,67-,68-,69-,70-,71-,72-,73-,74-,75-,76-,77-,78-,79-,80-,81-,82-,86-,87-,88-/m0/s1

InChI key

ZUBDGKVDJUIMQQ-ZTNLKOGPSA-N

Gene Information

human ... EDN1(1906)

General description

Endothelin-1 (ET-1) is synthesized and secreted by activated endothelial cells, and is a peptide with potent vasoconstrictor activity. It is a marker for activated or dysfunctional endothelial cells.[1] This peptide is one of the three isopeptides belonging to the endothelin family. It is also released by cardiomyocytes.[2]

Application

Endothelin-1 (ET-1) has been used to study the production and detection of H2O2 and production of NO in the presence of ET-1,[3] for studying the effects of ET-1 on the eNOS/inducible NOS (iNOS)/caveolin-1 expression, NO production, isoform specific PKC translocation, and PKC activity in human saphenous vein endothelial cells (HSVECs).[4]

Biochem/physiol Actions

Endothelin is a potent agonist for ETA and ETB endothelin receptors.
Endothelin-1 (ET-1) is a vasoconstrictor, which is increased in plasma and explants of patients with uterine leiomyomas. Thus, uterine fibroids are linked to increased ET-1 levels and hypertension, which is worsened by hypoxic condtions.[1] ET-1 RNAi and Endostar are capable of preventing the progression and invasiveness of PC-3 (prostate cancer) cells.[5] The plasma levels of this peptide are increased in patients with atrial septal defect (ASD), and these levels might help in diagnosis of closure defects.[2] The plasma level of ET-1 is also higher in patients with end-stage renal disease (ESRD), and this is linked to gradual worsening of pulmonary function tests.[6]
Potent agonist for ETA and ETB endothelin receptors. Induces the production of hypoxia-inducible factor 1α, and, thus, the production of VEGF.

Other Notes

A synthetic peptide with the sequence of human and porcine Endothelin 1.
Lyophilized from 0.1% TFA in H2O
The biological source is human endothelin, which shares the same sequence

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
WH0001906M1SRP3025E9012
technique(s)

ELISA: suitable

technique(s)

immunoprecipitation (IP): suitable, indirect ELISA: suitable, western blot: 1-5 μg/mL

technique(s)

cell culture | mammalian: suitable

technique(s)

-

assay

≥97% (HPLC)

assay

-

assay

≥95% (HPLC), ≥95% (SDS-PAGE)

assay

≥97% (HPLC)

biological source

human

biological source

mouse

biological source

human

biological source

-

form

powder

form

buffered aqueous solution

form

lyophilized

form

powder

solubility

1% acetic acid: >1 mg/mL, H2O: >1 mg/mL

solubility

-

solubility

-

solubility

-

Gene Information

human ... EDN1(1906)

Gene Information

human ... EDN1(1906)

Gene Information

human ... DLL1(28514)

Gene Information

human ... EDN2(1907)


Still not finding the right product?

Explore all of our products under Endothelin 1


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. What is the storage information for the reconstituted product? How long are aliquots stable at -20C?

    1 answer
    1. The reconstituted product has a limited lifetime in solution. Long-term storage in solution should be avoided.

      Helpful?

Reviews

No rating value

Active Filters