Skip to Content
Merck
All Photos(1)

Key Documents

E4660

Sigma-Aldrich

Eupatorin

≥97% (HPLC)

Synonym(s):

3′,5-Dihydroxy-4′,6,7-trimethoxyflavone, 5-Hydroxy-2-(3-hydroxy-4-methoxyphenyl)-6,7-dimethoxy-4H-1-benzopyran-4-one, 6-Methoxyluteolin 4′,7-dimethyl ether, NSC 106402

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H16O7
CAS Number:
Molecular Weight:
344.32
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

powder

color

white to beige

solubility

DMSO: >10 mg/mL

storage temp.

room temp

SMILES string

COc1ccc(cc1O)C2=CC(=O)c3c(O)c(OC)c(OC)cc3O2

InChI

1S/C18H16O7/c1-22-12-5-4-9(6-10(12)19)13-7-11(20)16-14(25-13)8-15(23-2)18(24-3)17(16)21/h4-8,19,21H,1-3H3

InChI key

KLAOKWJLUQKWIF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Eupatorin has been used as reference standard for characterization of Artemisia extracts using mass spectrometry (MS).

Biochem/physiol Actions

Eupatorin acts as an antiproliferative in cells expressing the CYP1A- family. It induces G2/M block follow by apoptosis in cells expressing the CYP1A- family. It also functions as an anti-inflammatory.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Anna-Leena Salmela et al.
Experimental cell research, 318(5), 578-592 (2012-01-10)
The spindle assembly checkpoint (SAC) is a conserved mechanism that ensures the fidelity of chromosome distribution in mitosis by preventing anaphase onset until the correct bipolar microtubule-kinetochore attachments are formed. Errors in SAC function may contribute to tumorigenesis by inducing
Yan Pan et al.
Journal of ethnopharmacology, 133(2), 881-887 (2010-11-26)
Andrographis paniculata (AP), Centella asiatica (CA) and Orthosiphon stamineus (OS) are three popular herbs traditionally used worldwide. AP is known for the treatment of infections and diabetes and CA is good for wound healing and healthy skin while OS is
I M Lyckander et al.
Prostaglandins, leukotrienes, and essential fatty acids, 54(4), 239-246 (1996-04-01)
15-Lipoxygenase from soybeans is inactivated by bubbling air through enzyme solutions. This inactivation is prevented by flavonoids from the East Asian medicinal plant Orthosiphon spicatus, previously found to be inhibitors of the enzyme. 5,7,4'-Trimethylapigenin, eupatorin and 5,7,3',4'-tetramethylluteolin show the strongest
Yit Hong Loon et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 816(1-2), 161-166 (2005-01-25)
A simple liquid chromatographic method was developed for the simultaneous determination of flavonoids from Orthosiphon stamineus Benth, namely sinensitin, eupatorin and 3'-hydroxy-5,6,7,4'-tetramethoxyflavone, in plasma. Prior to analysis, the flavonoids and the internal standard (naproxen) were extracted from plasma samples using
Iva Dolečková et al.
Fitoterapia, 83(6), 1000-1007 (2012-06-16)
Flavone eupatorin is one of the constituents of Orthosiphon stamineus, a medicinal herb used in folk medicine in South East Asia for treatment of various disorders. In our study, we investigated the antiproliferative properties of a chloroform extract of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service