Skip to Content
Merck
All Photos(2)

Key Documents

E2886

Sigma-Aldrich

ExtrAvidin®−Peroxidase

buffered aqueous solution

Synonym(s):

ExtrAvidin HRP

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46
Pricing and availability is not currently available.

conjugate

peroxidase conjugate

Quality Level

form

buffered aqueous solution

technique(s)

direct ELISA: 1:2,000
dot blot: 1:4,000
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100

shipped in

wet ice

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Related Categories

Specificity

The product is found to be reactive with Anti-Avidin, Catalog Number A5170, and non-reactive with Anti-Streptavidin, Catalog Number S6390, by immunoelectrophoresis.

Application

ExtrAvidin®-Peroxidase is suitable for the following applications:
  • Dot blot at a minimum dilution of 1:4,000
  • Immunohistochemistry (using formalin-fixed, paraffin-embedded sections at a dilution of 1:100)[1][2][3][4][5][6]
  • Direct ELISA at a minimum dilution of 1:2,000[7][8]
It has also been used for:
  • Immunoprecipitation and Immunobloting[9]
  • Ra2I solid-phase binding assays[10]
  • Southwestern blotting[11]
  • Western blotting[11][12][13]
  • Immunocytochemistry[14]

Biochem/physiol Actions

Avidin is a tetrameric or dimeric biotin-binding protein produced in the oviducts of birds, reptiles and amphibians and deposited in the whites of their eggs.[15] It consists of four high affinity binding sites for biotin. ExtrAvidin® is prepared from egg white avidin. It is a modified form of affinity purified avidin with high specific activity of avidin and low background staining of streptavidin. It is a biotin binding protein produced by the bacteria Streptomyces avidinii.[16] ExtrAvidin-Peroxidase may be used in conjunction with biotin in different techniques.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 0.05% MIT

Preparation Note

Affinity purified protein

Storage and Stability

Store at 2-8 °C. Do Not Freeze. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use.

Legal Information

ExtrAvidin is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohamed S Benleulmi et al.
Retrovirology, 14(1), 54-54 (2017-11-29)
Stable insertion of the retroviral DNA genome into host chromatin requires the functional association between the intasome (integrase·viral DNA complex) and the nucleosome. The data from the literature suggest that direct protein-protein contacts between integrase and histones may be involved
Stefan Lüth et al.
The Journal of clinical investigation, 118(10), 3403-3410 (2008-09-20)
Tregs are important mediators of immune tolerance to self antigens, and it has been suggested that Treg inactivation may cause autoimmune disease. Therefore, immunotherapy approaches that aim to restore or expand autoantigen-specific Treg activity might be beneficial for the treatment
H Kaur et al.
Scandinavian journal of immunology, 62(4), 361-370 (2005-10-29)
The present study was designed to examine the binding and signalling effects of single base and CpG dinucleotide phosphodiester (Po) oligodeoxynucleotides (ODN) on the human natural killer (NK)-like cell line (YT-INDY). Single base Po ODN composed of 20-mers of guanosine
D Tuckwell et al.
Journal of cell science, 108 ( Pt 4), 1629-1637 (1995-04-01)
Integrins alpha 1 beta 1 and alpha 2 beta 1 are major cellular receptors for collagens. The alpha 1 and alpha 2 subunits contain a approximately 200 amino acid inserted domain (I-domain) in their N-terminal region and, because of the
C I Martin et al.
The Journal of experimental biology, 208(Pt 11), 2083-2090 (2005-05-26)
Rainbow trout Oncorhynchus mykiss Walbaum were exercised at 0.8 and 1.6 body lengths s(-1) for 18 h a day over a 30 day period. Exercise resulted in a 24-30% increase in the average cross-sectional area of fast muscle fibres relative

Protocols

Use this protocol to for the entire immunohistochemistry (IHC) procedure through staining and visualization of specific antigens in paraffin-embedded tissue sections.

Use this protocol to for the entire immunohistochemistry (IHC) procedure through staining and visualization of specific antigens in paraffin-embedded tissue sections.

Use this protocol to for the entire immunohistochemistry (IHC) procedure through staining and visualization of specific antigens in paraffin-embedded tissue sections.

Use this protocol to for the entire immunohistochemistry (IHC) procedure through staining and visualization of specific antigens in paraffin-embedded tissue sections.

Questions

  1. Biotinylated protein을 detection하기 위하여 ExtrAvidin-peroxidase를 ELISA에 사용하고자하는데, well blocking solution으로 casein이나 Nonfat Dry Milk 등을 사용해도 괜찮을까요?

    1 answer
    1. Yes, casein or nonfat dry milk may be used as a blocking solution. Please see the link below to review the product datasheet:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/370/767/e2886dat-ms.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service