Skip to Content
Merck
All Photos(1)

Documents

D4293

Sigma-Aldrich

3,3′-Diaminobenzidine

SIGMAFAST, chromogenic, tablet

Synonym(s):

DAB tablets

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

product name

SIGMAFAST 3,3′-Diaminobenzidine tablets, tablet, To prepare 5 mL

form

tablet

storage temp.

−20°C

SMILES string

Nc1ccc(cc1N)-c2ccc(N)c(N)c2

InChI

1S/C12H14N4/c13-9-3-1-7(5-11(9)15)8-2-4-10(14)12(16)6-8/h1-6H,13-16H2

InChI key

HSTOKWSFWGCZMH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

SIGMAFAST 3,3′-Diaminobenzidine tablets has been used as a chromogen for visualization in immunohistochemistry of gonad tissue, thymus, dental chronic periapical lesions and gall bladder sections.
Developed for use in immunohistology and immunoblotting as a precipitating substrate for the localization of peroxidase activity. DAB is the immunohistology substrate of choice as it produces an intense brown-black stain which is resistant to alcohol. Slides stained with DAB may be dehydrated, mounted in resinous media, and stored for future reference. SIGMAFAST DAB Tablets require no additional buffers or steps to prepare an active substrate solution. Tablets are individually packaged in foil packets

Reconstitution

Each tablet set dissolved in 5 mL deionized water yields a ready-to-use buffered solution containing DAB and urea hydrogen peroxide.

Legal Information

SIGMAFAST is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Eye Dam. 1 - Muta. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Expression of pluripotent stem cell markers in the human fetal testis
Kerr CL, et al.
Stem Cells, 26(2), 412-421 (2008)
Receptor activator NFkappaB-ligand and osteoprotegerin protein expression in human periapical cysts and granulomas
Menezes R, et al.
Oral Surgery, Oral Medicine, Oral Pathology, Oral Radiology, and Endodontics, 102(3), 404-409 (2006)
Increased Bax/Bcl-2 ratio up-regulates caspase-3 and increases apoptosis in the thymus of patients with myasthenia gravis
Salakou S, et al.
In Vivo, 21(1), 123-132 (2007)
Anna Schildt et al.
Journal of Parkinson's disease, 9(1), 121-139 (2018-11-20)
Leucine-rich repeat kinase 2 (LRRK2) mutations are the most common genetic risk factor for Parkinson's disease (PD). While the corresponding pathogenic mechanisms remain largely unknown, LRRK2 has been implicated in the immune system. To assess whether LRRK2 mutations alter the
Interstitial cells of Cajal are present in human extrahepatic bile ducts
Ahmadi O, et al.
Journal of Gastroenterology and Hepatology, 25(2), 277-285 (2010)

Articles

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service