Skip to Content
Merck

Skip To

D9515

Driselase from Basidiomycetes sp.

powder, Protein ≥10 % by biuret

greener alternative

Greener Alternative

Synonym(s):

cellulase, laminarinase, xylanase

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352204
EC Number:
286-055-3
NACRES:
NA.54
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


conjugate

conjugate (Glucosaminoglycan)

Quality Segment

form

powder

composition

Protein, ≥10% biuret

greener alternative product characteristics

Waste Prevention
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

shipped in

wet ice

storage temp.

−20°C

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency and waste prevention when used in cellulosic ethanol research. For more information see the article in biofiles and Enzymes for Alternative Energy Research.

Application

The enzyme from Sigma has been used as a control while testing the ability of p-coumaroyl esterase to release p-coumaroyl and feruloyl groups from intact cell walls. It has also been used in the protoplast preparation from mycelia during a study to investigate the pathogenicity of Cochliobolus carbonum on Maize.
Driselase from Basidiomycetes has been used in a study to assess the digestion by fungal glycanases of arabinoxylans with different feruloylated side-chains. Driselase from Basidiomycetes has also been used in a study to investigate the purification, characterization, and mode of action of a rhamnogalacturonan hydrolase.

Biochem/physiol Actions

Driselase is a cell wall degrading enzyme that contains cellulase, hemicellulase, pectinase etc. Therefore, it is very effective in removing plant cell walls to make protoplasts.

Other Notes

Crude powder containing laminarinase, xylanase and cellulase.

Legal Information

Driselase is a trademark of ASKA Animal Health Co. Ltd.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A7130H2125C8546
description

powder, Protein ≥10 % by biuret

description

Type II-B, 20-80 units/mg protein (biuret)

description

powder, 0.3-3.0 unit/mg solid (using a β-galactose dehydrogenase system and locust bean gum as substrate)

description

lyophilized powder, ≥1 unit/mg solid

form

powder

form

powder

form

powder

form

lyophilized powder

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

2-8°C

shipped in

wet ice

shipped in

wet ice

shipped in

-

shipped in

-

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

sustainability

Greener Alternative Product

sustainability

Greener Alternative Product

sustainability

Greener Alternative Product

sustainability

Greener Alternative Product

composition

Protein, ≥10% biuret

composition

-

composition

-

composition

-


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1

Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters