
Dantrolene sodium salt

Synonym(s):
1-[(5-(p-Nitrophenyl)furfurylidene)amino]hydantoin sodium salt, Dantrolene, Sodium Salt
Empirical Formula (Hill Notation):
C14H9N4O5 · Na
CAS Number:
Molecular Weight:
336.23
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Research Area
Quality Segment
Showing 1-2 of 2