Skip to Content
Merck

Skip To

D8894

Dioleoylglycerol

≥99%, mixture of 1,3- and 1,2-isomers, liquid

Synonym(s):

Diolein, Glycerol-1,2- and -1,3-dioleate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C39H72O5
CAS Number:
Molecular Weight:
620.99
PubChem Substance ID:
UNSPSC Code:
51191904
MDL number:
NACRES:
NA.25
EC Number:
247-144-2
Beilstein/REAXYS Number:
1730341
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥99%

form

liquid

composition

mixture of 1,3- and 1,2-isomers

lipid type

neutral glycerides

storage temp.

−20°C

SMILES string

CCCCCCCC\C=C\CCCCCCCC(=O)OCC(O)COC(=O)CCCCCCC\C=C/CCCCCCCC.CCCCCCCC\C=C/CCCCCCCC(=O)OCC(CO)OC(=O)CCCCCCC\C=C\CCCCCCCC

InChI

1S/2C39H72O5/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-38(41)43-35-37(40)36-44-39(42)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2;1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-38(41)43-36-37(35-40)44-39(42)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h2*17-20,37,40H,3-16,21-36H2,1-2H3/b2*19-17-,20-18+

InChI key

WSCSXJUESYNIOA-SGKMRKLKSA-N

Packaging

Sealed ampule.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
723703741086709077
assay

≥99%

assay

95% (CP)

assay

97% (CP)

assay

95% (CP)

form

liquid

form

solid

form

liquid

form

liquid

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

composition

mixture of 1,3- and 1,2-isomers

composition

-

composition

-

composition

-

Quality Level

200

Quality Level

-

Quality Level

-

Quality Level

-

lipid type

neutral glycerides

lipid type

-

lipid type

-

lipid type

-


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters