Skip to Content
Merck
All Photos(1)

Key Documents

D7696

Sigma-Aldrich

Demethoxycurcumin

≥98% (HPLC), powder, LPS-induced nitric oxide (NO) production inhibitor

Synonym(s):

(E,E)-1-(4-Hydroxy-3-methoxyphenyl)-7-(4-hydroxyphenyl)-1,6-heptadiene-3,5-dione, Curcumin II, Desmethoxycurcumin, Monodemethoxycurcumin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H18O5
CAS Number:
Molecular Weight:
338.35
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

Demethoxycurcumin, ≥98% (HPLC)

Assay

≥98% (HPLC)

form

powder

storage condition

protect from light

color

orange

solubility

DMSO: ≥10 mg/mL

storage temp.

2-8°C

SMILES string

COc1cc(\C=C\C(=O)CC(=O)\C=C\c2ccc(O)cc2)ccc1O

InChI

1S/C20H18O5/c1-25-20-12-15(6-11-19(20)24)5-10-18(23)13-17(22)9-4-14-2-7-16(21)8-3-14/h2-12,21,24H,13H2,1H3/b9-4+,10-5+

InChI key

HJTVQHVGMGKONQ-LUZURFALSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Demethoxycurcumin (DMC) is a derivative or curcumin, and represents one of the major active components of curcumin products isolated from Curcumae sp. DMC inhibits LPS-induced nitric oxide (NO) production, and expression of iNOS and COX2 in RAW264.7 cells by blocking NF-kB activation. DMC also inhibits NF-kB dependent iNOS, TNFα and IL-1β expression in LPS-treated rat microglial cells. DMC suppresses expression of MMPs and ICAM-1 in MDA-MB-231 human breast cancer cells by inhibition of NF-kB.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hye Jin Kim et al.
Phytochemical analysis : PCA, 20(5), 372-377 (2009-06-18)
The new ion source technique, direct analysis in real time (DART) atmospheric pressure ionisation, allows high resolution mass measurements of gas, liquid and solid samples. As DART-MS produces [M + H](+) molecular ions of most compounds, relatively simple and clear
T Ahmed et al.
Neuroscience, 169(3), 1296-1306 (2010-06-12)
Alzheimer's disease (AD) is a neurodegenerative disease. There are a limited number of therapeutic options available for the treatment of AD. Curcuminoids (a mixture of bisdemethoxycurcumin, demethoxycurcumin and curcumin) is the main chemical constituent found in turmeric, a well known
Supachai Yodkeeree et al.
European journal of pharmacology, 627(1-3), 8-15 (2009-10-13)
Demethoxycurcumin (DMC) is one of the main active compounds of curcuminoids found in turmeric powder, which is used as a spice in Asian cooking and traditional medicine. Recent studies reveal that DMC has several biological activities including anti-inflammation and anti-cancer
Lijia Zhang et al.
International immunopharmacology, 10(3), 331-338 (2009-12-19)
Our previous report has showed that demethoxycurcumin (DMC), a natural derivative of curcumin (Cur), exhibited stronger inhibitory activity on nitric oxide (NO) and tumor necrosis factor-alpha (TNF-alpha) production compared with Cur in lipopolysaccharide (LPS) activated rat primary microglia. In the
Mathanaporn Boonrao et al.
Archives of pharmacal research, 33(7), 989-998 (2010-07-28)
Matrix metalloproteinase-3 (MMP-3) is a key enzyme with important implications in the invasion and metastasis of breast cancer cells. Curcumin (Cur), demethoxycurcumin (DMC), and bisdemethoxycurcumin (BDMC) are major forms of curcuminoids found in turmeric powder with reported anticancer activity. This

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service