Skip to Content
Merck
All Photos(2)

Key Documents

90593

Supelco

Demethoxycurcumin

analytical standard

Synonym(s):

(E,E)-1-(4-Hydroxy-3-methoxyphenyl)-7-(4-hydroxyphenyl)-1,6-heptadiene-3,5-dione, Curcumin II, Desmethoxycurcumin, Monodemethoxycurcumin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H18O5
CAS Number:
Molecular Weight:
338.35
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

SMILES string

COc1cc(\C=C\C(=O)CC(=O)\C=C\c2ccc(O)cc2)ccc1O

InChI

1S/C20H18O5/c1-25-20-12-15(6-11-19(20)24)5-10-18(23)13-17(22)9-4-14-2-7-16(21)8-3-14/h2-12,21,24H,13H2,1H3/b9-4+,10-5+

InChI key

HJTVQHVGMGKONQ-LUZURFALSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

This compound is commonly found in plants of the genus: curcuma

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Lijia Zhang et al.
International immunopharmacology, 10(3), 331-338 (2009-12-19)
Our previous report has showed that demethoxycurcumin (DMC), a natural derivative of curcumin (Cur), exhibited stronger inhibitory activity on nitric oxide (NO) and tumor necrosis factor-alpha (TNF-alpha) production compared with Cur in lipopolysaccharide (LPS) activated rat primary microglia. In the
Hye Jin Kim et al.
Phytochemical analysis : PCA, 20(5), 372-377 (2009-06-18)
The new ion source technique, direct analysis in real time (DART) atmospheric pressure ionisation, allows high resolution mass measurements of gas, liquid and solid samples. As DART-MS produces [M + H](+) molecular ions of most compounds, relatively simple and clear
Supachai Yodkeeree et al.
European journal of pharmacology, 627(1-3), 8-15 (2009-10-13)
Demethoxycurcumin (DMC) is one of the main active compounds of curcuminoids found in turmeric powder, which is used as a spice in Asian cooking and traditional medicine. Recent studies reveal that DMC has several biological activities including anti-inflammation and anti-cancer
Hongying Liu et al.
Neuroscience letters, 485(2), 83-88 (2010-08-24)
The hallmark of Alzheimer's disease (AD) is the accumulation of β-amyloid protein (Aβ). Aβ is generated from the β-amyloid precursor protein (APP) through the proteolysis of β-site APP cleaving enzyme 1 (BACE1) and γ-secretase. Aβ(42) isoform is more easily aggregate
Mathanaporn Boonrao et al.
Archives of pharmacal research, 33(7), 989-998 (2010-07-28)
Matrix metalloproteinase-3 (MMP-3) is a key enzyme with important implications in the invasion and metastasis of breast cancer cells. Curcumin (Cur), demethoxycurcumin (DMC), and bisdemethoxycurcumin (BDMC) are major forms of curcuminoids found in turmeric powder with reported anticancer activity. This

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service