Skip to Content
Merck
All Photos(1)

Key Documents

D3446

Sigma-Aldrich

Dipeptidyl Peptidase IV human

recombinant, expressed in Sf9 cells

Synonym(s):

CD26, DPPIV, Dipeptidyl aminopeptidase IV, Glycoprotein GP110

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

recombinant

expressed in Sf9 cells

Quality Level

form

solution

specific activity

≥4,000 units/μg protein

mol wt

124 kDa

concentration

≥0.01 mg/mL

UniProt accession no.

relevant disease(s)

cancer (lymphoma, prostate and colon cancer)

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... DPP4(1803)

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Dipeptidyl Peptidase IV (DPP4) is mapped to human chromosome 2q24.2. It comprises the N-glycosylation sites located in the β-propeller domain.

Application

Dipeptidyl Peptidase IV human has been used in DPP-IV inhibitory activity assay of quinoa protein concentrate digest.
Human dipeptidyl peptidase IV has been used in a study to assess the interactive hemodynamic effects of its inhibition as well as the angiotensin-converting enzyme inhibition. Human dipeptidyl peptidase IV has also been used in a study to identify and characterize the 100 kDa High Molecular Weight Hymenoptera Venom Allergens Api m 5 and Ves v 3.

Biochem/physiol Actions

Dipeptidyl Peptidase IV (DPP4) mediates the activation of T-cells. It is also a potential biomarker for lymphoma, thyroid, prostate and colon cancer.
Native DPPIV is a ubiquitous type II transmembrane glycoprotein and a serine protease of the S9 prolyl-oligopeptidase family. In vivo, it is synthesized with a signal peptide, which functions as the membrane anchoring domain. There is an 88% sequence homology between the human and porcine kidney enzymes. Both exist as homodimers with a subunit molecular weight of ~30 kDa. The high mannose 100 kDa DPPIV precursor is processed in the Golgi to yield a 124 kDa heavily N-and O-linked mature glycoprotein. It is then sorted to the apical membrane through the concerted action of both N- and O-linked glycans and its association with lipid microdomains. The porcine enzyme contains 18.3% carbohydrates, which the glycan composition is 0.9% fucose, 3.4% mannose, 5.1% galactose, 8.2% glucosamine, and 0.7% sialic acid. DPPIV is highly expressed on endothelial cells, epithelial cells, and lymphocytes. It is also present in plasma in its soluble form.

Unit Definition

One unit will hydrolyze 1.0 picomole of Ala-Pro-AMC per minute at pH 7.5 at 25 deg °C

Physical form

Supplied as a solution in 45 mM Tris-HCl, pH 8.0, 124 mM NaCl, 2.4 mM KCl, 225 mM imidazole and 10% glycerol.

Other Notes

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Release of dipeptidyl peptidase IV, alpha-amylase and alpha-glucosidase inhibitory peptides from quinoa (Chenopodium quinoa Willd.) during in vitro simulated gastrointestinal digestion
Vilcacundo R, et al.
Journal of functional foods, 35, 531-539 (2017)
N-linked glycosylation of dipeptidyl peptidase IV (CD26): Effects on enzyme activity, homodimer formation, and adenosine deaminase binding
Aertgeerts K, et al.
Protein Science, 13(1), 145-154 (2004)
Unravelling the immunological roles of dipeptidyl peptidase 4 (DPP4) activity and/or structure homologue (DASH) proteins
Wagner L, et al.
Clinical and Experimental Immunology, 184(3), 265-283 (2016)
Annis Marney et al.
Hypertension (Dallas, Tex. : 1979), 56(4), 728-733 (2010-08-04)
Dipeptidyl peptidase-IV inhibitors improve glucose homeostasis in type 2 diabetics by inhibiting degradation of the incretin hormones. Dipeptidyl peptidase-IV inhibition also prevents the breakdown of the vasoconstrictor neuropeptide Y and, when angiotensin-converting enzyme (ACE) is inhibited, substance P. This study
Simon Blank et al.
Journal of immunology (Baltimore, Md. : 1950), 184(9), 5403-5413 (2010-03-30)
Insect stings can cause life-threatening IgE-mediated anaphylactic reactions in venom-allergic patients. Although several compounds have already been described as venom allergens, prominent allergen candidates especially in the higher m.w. range have still remained elusive. Tandem mass spectrometry-based sequencing assigned a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service