Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Dipeptidyl Peptidase IV human
Synonym(s):
CD26, DPPIV, Dipeptidyl aminopeptidase IV, Glycoprotein GP110
Products
Product Category
proteases (1)
Brand
Sigma-Aldrich (2)
Gene Alias
Physical Form
Recombinant Host
Quality Segment
Relevant Disease
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
D4943
Dipeptidyl Peptidase IV human,
recombinant, expressed in baculovirus infected
Sf
9 cells, pkg of ≥1.0 units/vial, ≥10 units/mg protein
Expand
View Pricing
D3446
Dipeptidyl Peptidase IV human,
recombinant, expressed in
Sf
9 cells
Expand
View Pricing