Skip to Content
Merck

Skip To

O4876

Oligomycin

from Streptomyces diastatochromogenes, ≥90% total oligomycins basis (HPLC), powder, antibiotic

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Beilstein/REAXYS Number:
7615308
MDL number:
Assay:
≥90% total oligomycins basis (HPLC)
Form:
powder
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Oligomycin from Streptomyces diastatochromogenes, ≥90% total oligomycins basis (HPLC)

Quality Segment

assay

≥90% total oligomycins basis (HPLC)

form

powder

antibiotic activity spectrum

fungi

mode of action

enzyme | inhibits

storage temp.

−20°C

InChI

1S/C45H74O11/c1-12-34-17-15-13-14-16-27(4)42(51)44(11,53)43(52)32(9)40(50)31(8)39(49)30(7)38(48)26(3)18-21-37(47)54-41-29(6)35(20-19-34)55-45(33(41)10)23-22-25(2)36(56-45)24-28(5)46/h13-15,17-18,21,25-36,38,40-42,46,48,50-51,53H,12,16,19-20,22-24H2,1-11H3/b14-13+,17-15-,21-18+

InChI key

MNULEGDCPYONBU-ZUSSGZTJSA-N

General description

Chemical structure: macrolide

Application

Oligomycin from Streptomyces diastatochromogenes is used:
  • in the inhibition of mitochondrial ATP production Leigh syndrome (LS) cells.
  • in ATP-linked respiration and maximal capacity respiration assay in astrocytes.
  • in measurement of oxygen consumption rate (OCR) and extracellular acidification rates in breast cancer cell lines.

Biochem/physiol Actions

Oligomycin belongs to the macrolide group of antibiotics. It inhibits mitochondrial ATP-synthase enzyme and phosphoryl group transfer. Oligomycin interacts with ATP synthase subunit-c in the Fo portion and impairs proton transport.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
75351B1793R0395
form

powder

form

powder

form

film

form

powder

assay

≥90% total oligomycins basis (HPLC)

assay

≥99% (HPLC)

assay

≥90% (HPLC)

assay

≥95% (HPLC)

Quality Level

200

Quality Level

200

Quality Level

300

Quality Level

300

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

antibiotic activity spectrum

fungi

antibiotic activity spectrum

fungi

antibiotic activity spectrum

fungi

antibiotic activity spectrum

fungi, yeast

mode of action

enzyme | inhibits

mode of action

enzyme | inhibits

mode of action

DNA synthesis | interferes, enzyme | inhibits

mode of action

enzyme | inhibits, protein synthesis | interferes


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. What is the molecular weight of this compound mixture

    1 answer
    1. This product is a mixture of oligomycin's A, B, and C. The molecular weight of each is as follows: :
      Oligomycin A = 791 g/mol (approximately 65%)
      Oligomycin B = 805.1 g/mol (amount not determined)
      Oligomycin C = 775.1 g/mol (amount not determined)

      Please see the link below to review the product datasheet:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/165/401/o4876pis.pdf

      Helpful?

Reviews

No rating value

Active Filters