Skip to Content
Merck
All Photos(1)

Key Documents

D3160

Sigma-Aldrich

Deoxyribonucleic acid from human placenta

buffered aqueous solution, sexed, male

Synonym(s):

DNA

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200

grade

for molecular biology

form

buffered aqueous solution

shipped in

dry ice

storage temp.

−20°C

InChI

1S/C15H31N3O13P2/c16-13-1-7(20)11(28-13)5-25-32(21,22)31-9-3-15(18)29-12(9)6-26-33(23,24)30-8-2-14(17)27-10(8)4-19/h7-15,19-20H,1-6,16-18H2,(H,21,22)(H,23,24)

InChI key

AWBASQCACWFTGD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Human placental DNA is isolated from donor placenta, but will contain some maternal DNA. The DNA fragments are sonicated to produce fragments of consistent size.

Application

Deoxyribonucleic acid from human placenta is used as blocking agent in Southern hybridizations. It was used to compare the efficiency of DNA quantification methods.
For use in Southern hybridizations.

Components

DNA is supplied in a solution of 10mM Tris-HCl (pH 8.4), with 1mM EDTA.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karsten Nielsen et al.
Forensic science international. Genetics, 2(3), 226-230 (2008-12-17)
Six commercial preparations of human genomic DNA were quantified using five quantification methods: UV spectrometry, SYBR-Green dye staining, slot blot hybridization with the probe D17Z1, Quantifiler Human DNA Quantification kit and RB1 rt-PCR. All methods measured higher DNA concentrations than
Sriram Kosuri et al.
Nature methods, 11(5), 499-507 (2014-05-02)
For over 60 years, the synthetic production of new DNA sequences has helped researchers understand and engineer biology. Here we summarize methods and caveats for the de novo synthesis of DNA, with particular emphasis on recent technologies that allow for
Xin Cai et al.
Molecular cell, 54(2), 289-296 (2014-04-29)
The innate immune system deploys a variety of sensors to detect signs of infection. Nucleic acids represent a major class of pathogen signatures that can trigger robust immune responses. The presence of DNA in the cytoplasm of mammalian cells is
Giang Huong Nguyen et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(27), 9905-9910 (2014-06-25)
Bloom syndrome is a rare autosomal recessive disorder characterized by genetic instability and cancer predisposition, and caused by mutations in the gene encoding the Bloom syndrome, RecQ helicase-like (BLM) protein. To determine whether altered gene expression might be responsible for
Arnaud Obri et al.
Nature, 505(7485), 648-653 (2014-01-28)
H2A.Z is an essential histone variant implicated in the regulation of key nuclear events. However, the metazoan chaperones responsible for H2A.Z deposition and its removal from chromatin remain unknown. Here we report the identification and characterization of the human protein

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service