Skip to Content
Merck

Skip To

D2653

Dimethyl sulfoxide

Hybri-Max, 0.2 μm filtered, BioReagent, suitable for hybridoma, ≥99.7%, Ampoule

Synonym(s):

Dimethyl sulfoxide, DMSO

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
(CH3)2SO
CAS Number:
Molecular Weight:
78.13
UNSPSC Code:
12191502
Beilstein/REAXYS Number:
506008
MDL number:
NACRES:
NA.21
Assay:
≥99.7%
Grade:
Hybri-Max
Form:
liquid
Solubility:
H2O: miscible (completely)
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

Hybri-Max

Quality Segment

vapor density

2.7 (vs air)

vapor pressure

0.42 mmHg ( 20 °C)

sterility

0.2 μm filtered

product line

BioReagent

assay

≥99.7%

form

liquid

autoignition temp.

573 °F

expl. lim.

42 %, 63 °F

technique(s)

cell culture | hybridoma: suitable, cryopreservation: suitable

impurities

endotoxin, tested, ≤0.1% water

refractive index

n20/D 1.479 (lit.)

bp

189 °C (lit.)

mp

16-19 °C (lit.)

solubility

H2O: miscible (completely)

density

1.10 g/mL (lit.)

SMILES string

CS(C)=O

InChI

1S/C2H6OS/c1-4(2)3/h1-2H3

InChI key

IAZDPXIOMUYVGZ-UHFFFAOYSA-N

Application

DMSO is a polar aprotic solvent used in chemical reactions, in polymerase chain reactions (PCR) and as a cryoprotectant vitrification agent for the preservation of cells, tissues and organs. DMSO is used in cell freezing media to protect cells from ice crystal induced mechanical injury. It is used for frozen storage of primary, sub-cultured, and recombinant heteroploid and hybridoma cell lines; embryonic stem cells (ESC), and hematopoietic stem cells. DMSO is frequently used in the combinations with BSA or fetal bovine serum (FBS).

Packaging

5 mL and 10 mL in flame sealed ampules

Other Notes

This product is a Hybri-Max product. It is hybridoma tested and is assessed for suitability in cell freezing. This product is 0.2 micron filtered and tested for endotoxin levels.

Legal Information

Hybri-Max is a trademark of Sigma-Aldrich Co. LLC

Disclaimer

Supercools easily and remelts slowly at room temperature. Solidified product can be re-liquified by warming to room temperature without detriment to the product.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
D2650D2438D4540
description

Hybri-Max, 0.2 μm filtered, BioReagent, suitable for hybridoma, ≥99.7%, Ampoule

description

Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%

description

sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma

description

≥99.5% (GC), suitable for plant cell culture

form

liquid

form

liquid

form

liquid

form

liquid

solubility

H2O: miscible (completely)

solubility

H2O: miscible (completely)

solubility

H2O: miscible (completely)

solubility

H2O: miscible (completely)

assay

≥99.7%

assay

≥99.7%

assay

-

assay

≥99.5% (GC)

Quality Level

300

Quality Level

400

Quality Level

400

Quality Level

200

grade

Hybri-Max

grade

Hybri-Max

grade

BioPerformance Certified

grade

-

density

1.10 g/mL (lit.)

density

1.10 g/mL (lit.)

density

1.10 g/mL (lit.)

density

1.10 g/mL (lit.)


Still not finding the right product?

Explore all of our products under Dimethyl sulfoxide


Storage Class

10 - Combustible liquids

flash_point_f

188.6 °F - closed cup

flash_point_c

87 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library