Skip to Content
Merck
All Photos(2)

Key Documents

D0125

Sigma-Aldrich

Dansyl-L-alanine cyclohexylammonium salt

≥98% (TLC), suitable for protein expression

Synonym(s):

N-[[5-(Dimethylamino)-1-naphthalenyl]sulfonyl]- L-alanine cyclohexylammonium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H18N2O4S · C6H13N
CAS Number:
Molecular Weight:
421.55
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

product name

Dansyl-L-alanine cyclohexylammonium salt, fluorescent amino acid

Quality Level

Assay

≥98% (TLC)

form

powder

technique(s)

protein expression: suitable

storage temp.

−20°C

SMILES string

NC1CCCCC1.C[C@H](NS(=O)(=O)c2cccc3c(cccc23)N(C)C)C(O)=O

InChI

1S/C15H18N2O4S.C6H13N/c1-10(15(18)19)16-22(20,21)14-9-5-6-11-12(14)7-4-8-13(11)17(2)3;7-6-4-2-1-3-5-6/h4-10,16H,1-3H3,(H,18,19);6H,1-5,7H2/t10-;/m0./s1

InChI key

CZUBQNQWSOLSJH-PPHPATTJSA-N

Application

Alanine conjugated to the fluorophore 5-(dimethylamino)naphthalene-1-sulfonyl (dansyl). The unnatural amino acid has been incorporated into expressed proteins by use of a mutated aminoacyl-tRNA synthetase specific for dansylalanine.

Biochem/physiol Actions

Dansyl-L-alanine contains an N-terminal fluorophore (dansyl) making it useful as a fluorescent marker/probe. Dansylalanine can be incorporated into proteins by newly developing techniques that expand the genetic code.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A genetically encoded fluorescent amino acid.
Summerer, D., et al.
Proceedings of the National Academy of Sciences of the USA, 108, 9785-9789 (2006)
Wenyuan Wang et al.
Nature neuroscience, 10(8), 1063-1072 (2007-07-03)
Proteins participate in various biological processes and can be harnessed to probe and control biological events selectively and reproducibly, but the genetic code limits the building block to 20 common amino acids for protein manipulation in living cells. The genetic
Hyun Soo Lee et al.
Journal of the American Chemical Society, 131(36), 12921-12923 (2009-08-26)
Here, we report that the fluorescent amino acid, 3-(6-acetylnaphthalen-2-ylamino)-2-aminopropanoic acid (Anap), can be genetically incorporated into proteins in yeast with excellent selectivity and efficiency by means of an orthogonal tRNA/aminoacyl-tRNA synthetase pair. This small, environmentally sensitive fluorophore was site-specifically incorporated
Daniel Summerer et al.
Proceedings of the National Academy of Sciences of the United States of America, 103(26), 9785-9789 (2006-06-21)
The ability to introduce fluorophores selectively into proteins provides a powerful tool to study protein structure, dynamics, localization, and biomolecular interactions both in vitro and in vivo. Here, we report a strategy for the selective and efficient biosynthetic incorporation of
Esterification of an unnatural amino acid structurally deviating from canonical amino acids promotes its uptake and incorporation into proteins in mammalian cells.
Jeffrey K Takimoto et al.
Chembiochem : a European journal of chemical biology, 11(16), 2268-2272 (2010-09-28)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service