Skip to Content
Merck
All Photos(1)

Key Documents

CLS3656

Corning® 384 well microplate

clear polypropylene, round bottom, sterile, lid: no, pack of 25

Synonym(s):

384 multiwell plate, 384 well assay plate, 384 well general storage plate, 384 well microplate, 384 well microtiter plate, 384 well plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.13
material:
clear bottom
clear polypropylene
round bottom
square wells
size:
384 wells
Pricing and availability is not currently available.

material

clear bottom
clear polypropylene
round bottom
square wells

sterility

sterile

feature

lid: no
skirt
microplate for general storage and assays
plate format: 384 well standard

packaging

case of 100
pack of 25

manufacturer/tradename

Corning 3656

size

384 wells

well volume

95 μL

well working volume

15-70 μL

color

clear

suitability

suitable for (cell cuture)

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

  • Resistant to many common organic solvents (e.g., DMSO, ethanol, methanol)
  • Certified DNase- and RNase-free
  • Total well volume 95 μL; working volume 15-70 μL

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service