Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Corning
®
384 well microplate
Products
Product Category
plates (11)
Color
Manufacturer
Binding Type
Volume (µL)
Wells
Color
Package Size
Sterility
Filter Products
Showing 1-11 of 11
Compare
Product Number
Description
Pricing
CLS3701
Corning
®
384 well microplate,
Tissue Culture (TC)-treated surface, clear polystyrene, flat bottom, sterile, lid, case of 100
Expand
View Pricing
CLS3702
Corning
®
384 well microplate,
clear polystyrene, flat bottom, non-sterile, lid: no, case of 100
Expand
View Pricing
CLS3656
Corning
®
384 well microplate,
clear polypropylene, round bottom, sterile, lid: no, pack of 25
Expand
View Pricing
CLS3985
Corning
®
384 well microplate,
Tissue Culture (TC)-treated surface, black polystyrene, square, sterile, lid, case of 100
Expand
View Pricing
CLS3700
Corning
®
384 well microplate,
clear polystyrene, flat bottom, non-sterile, lid: no, case of 100
Expand
View Pricing
CLS3657
Corning
®
384 well microplate,
clear polypropylene, round bottom, non-sterile, lid: no, pack of 25
Expand
View Pricing
CLS3702BC
Corning
®
384 well microplate,
clear polystyrene, flat bottom, non-sterile, with generic barcode, lid: no, case of 100
Expand
View Pricing
CLS3658
Corning
®
384 well microplate,
black polypropylene, round bottom, non-sterile, lid: no, pack of 25
Expand
View Pricing
CLS4681
Corning
®
384 well microplate,
black cyclic olefin copolymer (COC) (COC), flat bottom, sterile, lid, pack of 20
Expand
View Pricing
CLS3847
Corning
®
384 well microplate,
clear polystyrene, flat bottom, non-sterile, lid, pack of 20 ea
Expand
View Pricing
CLS3985BC
Corning
®
384 well microplate,
Tissue Culture (TC)-treated surface, clear polystyrene, flat bottom, non-sterile, with generic barcode, lid, case of 50
Expand
View Pricing