CLS3575
Corning® NBS™ 384 well microplates
NBS™ (non-binding surface), black polystyrene, flat bottom, non-sterile, lid: no, pack of 50
Synonym(s):
384 multiwell plates, 384 well immunoassay plates, 384 well microplates, 384 well microtiter plates, 384 well plates, immunoassay plates
About This Item
flat bottom
flat bottom wells black polystyrene wells
polystyrene
square black wells
Recommended Products
material
black polystyrene
flat bottom
flat bottom wells black polystyrene wells
polystyrene
square black wells
sterility
non-sterile
non-sterile
feature
lid: no
lid: no
skirt
packaging
pack of 50
manufacturer/tradename
Corning 3575
maximum volume
112 μL
size
384 wells
wells
384 wells
working volume
80 μL
color
black
suitability
suitable for (cell cuture)
binding type
NBS™ (non-binding surface)
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
- Black wall plates have reduced autofluorescence, higher signal to background ratio and lower CV′s.
- Nonbinding (NBS™) surface is a Corning proprietary treatment technology used on polystyrene microplates to create a nonionic hydrophilic surface (polyethylene oxide-like) that minimizes molecular interactions
- Ideal for reducing protein and nucleic acid binding at low concentrations, and increasing assay signal to noise
Note: Suggested replacement for CLS3654
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service