Skip to Content
Merck

CLS3571

Corning® 384 well microplate, low flange

Tissue Culture (TC)-treated surface, black polystyrene, flat bottom, sterile, lid, pack of 10

Synonym(s):

384 multiwell plates, 384 well low volume plates, 384 well microplates, 384 well microtiter plates, 384 well plates, 384 well tissue culture treated plates

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.24
UNSPSC Code:
41121800
Material:
black bottom, black polystyrene , flat bottom , polystyrene , square black wells
Size:
384 wells
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

black bottom, black polystyrene , flat bottom , polystyrene , square black wells

sterility

sterile

feature

lid, skirt

packaging

pack of 10

manufacturer/tradename

Corning 3571

maximum volume

112 μL

size

384 wells

working volume

80 μL

color

black

suitability

suitable for (cell cuture)

binding type

Tissue Culture (TC)-treated surface

General description

Black wall plates have reduced autofluorescence, higher signal to background ratio and lower CV′s.
• Enhanced flatness provdes low intra- and inter- plate CV′s.
• Flat bottoms with 112μL total volume
• Conforms to standard microplate footprint and dimensions
• TC-treated for optimal cell attachment
• Nonreversable lids with condensation rings to reduce contamination
• Sterilized by gamma radiation and certified nonpyrogenic
• Ideal for today′s more sensitive assays and plate readers.

Legal Information

Corning is a registered trademark of Corning, Inc.


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library