CLS354233
Corning® Collagen IV
from mouse(Natural), pack of 1 mg
Synonym(s):
Collagen Type IV, Type IV Collagen
About This Item
Recommended Products
biological source
mouse (Natural)
form
liquid
packaging
pack of 1 mg
manufacturer/tradename
Corning 354233
technique(s)
cell attachment: suitable
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Source
Source material tested for HBsAG, for anti-HIV-1 and anti-HIV-2, and anti-HCV virus-1 and virus-2.
Quality
Electrophoretically homogeneous (SDS-PAGE)
Tested and found negative for bacteria, fungi, and mycoplasma
Preparation and Storage
Stable for at least three months at -70°C. Avoid multiple freeze-thaws.
Handling
Recommended concentration: 1-5 μg/cm² of growth surface depending on cell type
Formulation
Frozen in 0.05 M HCI
For Customers in California only: WARNING Please refer to Safety Data Sheet (SDS) Section 15 or Letter in Lieu for more information regarding California Proposition 65.
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8B - Non-combustible corrosive hazardous materials
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service