Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
NACRES:
NA.54
UNSPSC Code:
41116158
Pricing and availability is not currently available.
Quality Segment
General description
The Imprint Ultra Chromatin Optimization Kit provides reagents needed to optimize chromatin sonication/shearing conditions before performing ChIP. It is imperative that chromatin is completely sheared into fragments with a peak around 500 base pairs (bp) for ChIP-qPCR assays, and < 500 bp for ChIP-Seq and ChIP-chip, for maximum specificity and resolution, and to ensure low background. For higher resolution applications like genome-wide location analysis by ChIP-chip or ChIP-Seq, sonication of input chromatin to below 500 bp, with a peak around 250 bp, is critical.
Other Notes
Sufficient reagents are provided to perform three sets of chromatin sonication optimization experiments. Each set would include five different conditions for sonication to optimize the parameters. Reagents are provided for a total of 15 reactions, including preparation of cell lysates from 107 cells/reaction, nuclei lysis, sonication, cross-link reversal and DNA purification.
Legal Information
Imprint is a registered trademark of Merck KGaA, Darmstadt, Germany
1 of 1
This Item | |||
|---|---|---|---|
| Quality Level 200 | Quality Level 200 | Quality Level 200 | Quality Level 200 |
Kit Components Also Available Separately
Product No.
Description
SDS & Pricing
signalword
Danger
target_organs
Central nervous system
Storage Class
5.1A - Strongly oxidizing hazardous materials
flash_point_f
100.4 °F
flash_point_c
38 °C
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - ED ENV 1 - Eye Dam. 1 - Flam. Liq. 3 - Met. Corr. 1 - Ox. Liq. 1 - Resp. Sens. 1 - Skin Corr. 1A - STOT SE 3
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.









