Skip to Content
Merck
All Photos(1)

Documents

BeWo Cell Line human

86082803, human placenta, Epithelial

Synonym(s):

Be-Wo Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

BeWo Cell Line human, 86082803

biological source

human placenta

description

Human choriocarcinoma

growth mode

Adherent

karyotype

modal number =86 range 71-178

morphology

Epithelial

products

Gonadotrophin, lactogen and steroid hormones

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human choriocarcinoma

Cell Line Description

The first human, trophoblastic endocrine cell type to be maintained in continuous culture. It was initiated from a malignant gestational choriocarcinoma of the foetal placenta. The cells secrete placental hormones including chorionic gonadotrophin (hCG), polypeptide hormones, lactogen, estrogenic and progestational steroids, estrone, estradiol, estriol and progesterone. A high rate of hCG Synthesis has been shown. Cellular differentiation of this stem cell type occurs when hormonal precursors are added to the medium. Due to the high glycolytic activity the cells have to be sub-cultured or supplied with fresh media every 24-48 hours to prevent glucose exhaustion. BeWo has the same DNA profile as JEG3 (Sigma catalogue number 92120308); JEG3 was established by serial cloning of BeWo.

Application

Hormone studies
Human BeWo cell line has been used to study the effects of Δ-9-tetrahydrocannabinol (Δ9-THC) on the proliferation and gene transcription in the BeWo trophoblast cell line. It has also been used to study placental transfer of highly purified non-dioxin-like polychlorinated biphenyls in BeWo cells.

DNA Profile

STR-PCR Data: Amelogenin: X,Y
CSF1PO: 11,12
D13S317: 9,11
D16S539: 13,14
D5S818: 10,11
D7S820: 10,12
THO1: 9,9.3
TPOX: 8
vWA: 16

Culture Medium

Ham′s F12 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 i.e. seeding at 4x10,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37°C; medium change or subculture every 24-48 hours necessary.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service