Skip to Content
Merck
All Photos(1)

Key Documents

L929(NCTC)

85103115, mouse adipose tissue, Fibroblast

Synonym(s):

L (929) Cells, NCTC 929 Cells, NCTC clone 929 Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

L929(NCTC), 85103115

biological source

mouse adipose tissue

growth mode

Adherent

karyotype

Not specified

morphology

Fibroblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse C3H/An areolar and adipose tissue

Cell Line Description

Derived from normal subcutaneous connective tissue of an adult C3H strain mouse. The cells were treated with 20 methylcholanthrene and produced sarcomas when injected into C3H strain mice.

Culture Medium

DMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split semi-confluent cultures 1:4 to 1:6 i.e. seeding at 2-4x10,000 cells /cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service