Skip to Content
Merck

217356

Carboxypeptidase B, Porcine Pancreas

Synonym(s):

Carboxypeptidase B, Porcine Pancreas

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
EC Number:
UNSPSC Code:
12352202
NACRES:
NA.54
MDL number:
Specific activity:
≥100 units/mg protein
Biological source:
Porcine pancreas
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

Porcine pancreas

Quality Segment

form

lyophilized

specific activity

≥100 units/mg protein

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

pI 

6.0

solubility

water: 1 mg/mL

foreign activity

Carboxypeptidase A <1.0%, Chymotrypsin, none detected, Trypsin ≤5%

shipped in

ambient

storage temp.

−20°C

SMILES string

S(Cc3ccccc3)C(=O)C(NC(=O)C2N(CCC2)C(=O)C(NC(=O)C(NC(=O)CCC(=O)O)C)C)Cc1ccccc1

InChI

1S/C31H38N4O7S/c1-20(32-26(36)15-16-27(37)38)28(39)33-21(2)30(41)35-17-9-14-25(35)29(40)34-24(18-22-10-5-3-6-11-22)31(42)43-19-23-12-7-4-8-13-23/h3-8,10-13,20-21,24-25H,9,14-19H2,1-2H3,(H,32,36)(H,33,39)(H,34,40)(H,37,38)

InChI key

TWURVFFNODFJBJ-UHFFFAOYSA-N

General description

Native carboxypeptidase B catalyzes the release of C-terminal from basic amino acids, arginine, lysine, or ornithine from polypeptides. Inhibited by EDTA and other Zn2+ chelators. Has an optimal pH of 7.0-9.0 and a pI of 6.0.

Physical form

Lyophilized from 360 µM HEPES, 1.41 µM zinc chloride, 1.41 mg/ml Trehalose, pH 7.4.

Preparation Note

Following reconstitution aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Folk, J.E. 1970. Methods Enzymol.19, 504.
One unit is defined as the amount of enzyme that will hydrolyze 1.0 µmol of Hip-Arg per min at 25°C, pH 7.7.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

wgk

WGK 3

Storage Class

11 - Combustible Solids

flash_point_f

No data available

flash_point_c

No data available



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library