Skip to Content
Merck

Skip To

C8032

α-Casein Dephosphorylated from bovine milk

lyophilized powder

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.61
MDL number:
Form:
lyophilized powder
Assay:
≥70% (biuret)
Biological source:
bovine milk
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

bovine milk

Quality Segment

assay

≥70% (biuret)

form

lyophilized powder

technique(s)

surface plasmon resonance (SPR): suitable

storage temp.

−20°C

SMILES string

[P](=O)(OCC(NC(=O)C(NC(=O)C(N)Cc1ccccc1)CCC(=O)N)C(=O)NC(CCC(=O)O)C(=O)NC(CCC(=O)O)C(=O)NC(CCC(=O)N)C(=O)NC(CCC(=O)N)C(=O)NC(CCC(=O)N)C(=O)NC(C(O)C)C(=O)NC(CCC(=O)O)C(=O)NC(CC(=O)O)C(=O)NC(CCC(=O)O)C(=O)NC(CC(C)C)C(=O)NC(CCC(=O)N)C(=O)NC(CC(=O)O)C(=O)NC(C

InChI

1S/C81H125N22O39P/c1-36(2)31-50(76(132)94-43(15-24-57(87)108)71(127)101-52(34-64(120)121)78(134)98-49(81(137)138)11-7-8-30-82)99-72(128)47(19-28-61(114)115)95-77(133)51(33-63(118)119)100-73(129)48(20-29-62(116)117)97-80(136)65(37(3)104)103-75(131)44(16-25-58(88)109)92-68(124)42(14-23-56(86)107)90-67(123)41(13-22-55(85)106)91-69(125)45(17-26-59(110)111)93-70(126)46(18-27-60(112)113)96-79(135)53(35-142-143(139,140)141)102-74(130)40(12-21-54(84)105)89-66(122)39(83)32-38-9-5-4-6-10-38/h4-6,9-10,36-37,39-53,65,104H,7-8,11-35,82-83H2,1-3H3,(H2,84,105)(H2,85,106)(H2,86,107)(H2,87,108)(H2,88,109)(H,89,122)(H,90,123)(H,91,125)(H,92,124)(H,93,126)(H,94,132)(H,95,133)(H,96,135)(H,97,136)(H,98,134)(H,99,128)(H,100,129)(H,101,127)(H,102,130)(H,103,131)(H,110,111)(H,112,113)(H,114,115)(H,116,117)(H,118,119)(H,120,121)(H,137,138)(H2,139,140,141)

InChI key

BECPQYXYKAMYBN-UHFFFAOYSA-N

Gene Information

Application

α-Casein from bovine milk is an Immunoglobulin E (IgE)-binding epitope belonging to the Casein phosphoprotein family. It has been used in studies of children with insulin-dependent diabetes mellitus (IDDM), due to the strong cross-reactivity of α-casein antibodies with insulin. Studies of children displaying (IDDM) show higher levels of α-casein antibodies than control siblings, which may serve as a risk marker for IDDM.

Biochem/physiol Actions

Clusters of phosphoserine in casein chelate iron and reduce its bioavailability. Partial enzymatic digestion of casein unmasks phosphorylated residues to alkaline phosphatase, permitting dephosphorylation and increasing the bioavailability of iron.

Preparation Note

Enzymatically prepared from C6780.

Analysis Note

At least 80% dephosphorylated.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C9801C3400C5890
description

lyophilized powder

description

essentially salt-free, lyophilized powder

description

powder

description

suitable for substrate for protein kinase (after dephosphorylation), purified powder

assay

≥70% (biuret)

assay

≥90% (colorimetric)

assay

87-94% protein basis

assay

-

biological source

bovine milk

biological source

bovine milk

biological source

bovine milk

biological source

bovine milk

technique(s)

surface plasmon resonance (SPR): suitable

technique(s)

activity assay: suitable

technique(s)

activity assay: suitable, electrophoresis: suitable, immunocytochemistry: suitable

technique(s)

activity assay: suitable, electrophoresis: suitable, immunocytochemistry: suitable

form

lyophilized powder

form

essentially salt-free, lyophilized powder

form

powder

form

purified powder

Gene Information

bovine ... MACAS(100532647), MACASP(105753520)

Gene Information

bovine ... CSN1S1(282208), CSN1S2(282209), CSN2(281099), CSN3(281728)

Gene Information

bovine ... CSN1S1(282208), CSN1S2(282209), CSN2(281099), CSN3(281728)

Gene Information

bovine ... CSN1S1(282208), CSN1S2(282209), CSN2(281099), CSN3(281728)

storage temp.

−20°C

storage temp.

−20°C

storage temp.

-

storage temp.

-


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters