Skip to Content
Merck
All Photos(1)

Key Documents

C7738

Sigma-Aldrich

Anti-β-Catenin−Cy3 antibody, Mouse monoclonal

clone 15B8, purified from hybridoma cell culture

Synonym(s):

Monoclonal Anti-β-Catenin

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

CY3 conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

15B8, monoclonal

form

buffered aqueous solution

mol wt

antigen 94 kDa

species reactivity

chicken, bovine, human, canine

concentration

~1 mg/mL

technique(s)

direct immunofluorescence: 1:100 using methanol/acetone-fixed bovine MDBK/MCF-7 Cell Line

isotype

IgG1

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... CTNNB1(1499)

Looking for similar products? Visit Product Comparison Guide

General description

Anti-β-Catenin-Cy3 antibody, Mouse monoclonal (mouse IgG1 isotype) is derived from the 15B8 hybridoma, produced by fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with recombinant chicken catenin. β-catenin is 92 kDa. β-catenin protein is composed of a series of protein-protein interaction motifs that allow it to function as a scaffold. The N-terminus domain contains the binding site for β-catenin as well as phosphorylation sites recognized by Glycogen synthase kinase 3 beta (GSK3β) and the C-terminus contains the transcriptional activation domain and binding site for Teashirt.
The gene β-catenin (CTNNB1) is mapped to human chromosome 3p21. The protein localizes to the cytoplasm and the nucleus. β-catenin is also present at the centrosomes.

Specificity

Recognizes specifically mammalian and avian β-catenin. Does not cross-react with plakoglobin (γ-catenin).

Immunogen

recombinant chicken β-catenin.

Application

Anti-β-Catenin-Cy3 antibody has been used in
  • direct immunofluorescence
  • western blotting
  • staining
  • immunohistochemistry

Suitable for direct immunofluorescence at a dilution of 1:100 using methanol/acetone-fixed bovine MDBK/MCF-7 cells. The antiobody has been used for western blot analysis in mouse embryonic stem cells.

Biochem/physiol Actions

Catenin (cadherin-associated protein) β 1 also called β-catenin is encoded by the CTNNB1 gene in humans. It is a dual function protein that regulates the coordination of cell-cell adhesion and gene transcription. It is useful for differential diagnosis of osteoblastoma (OB) and osteosarcoma (OS). β-catenin signaling pathway may contribute to cholangiocarcinoma (CCA) cell proliferation. It may serve as a prognostic marker for CCA progression and provide a potential target for CCA therapy. Mutations in β-catenin are associated with ovarian carcinoma. In addition, mutations in β-catenin are linked to intellectual disability. NEK2 (Never in mitosis A-related kinase 2) binds and phosphorylates β-catenin, thereby regulating centrosome seperation. Focal adhesion protein, Kindlin-2, enhances WNT signaling by forming a transcriptional complex with β-catenin.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide.

Legal Information

Cy is distributed under license from Amersham Biosciences Limited.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tissue factor and thrombomodulin expression on endothelial cell-seeded collagen modules for tissue engineering
She M, et al.
Journal of Biomedical Materials Research Part A, 80(2), 497-504 (2007)
Self-regulation of functional pathways by motifs inside the disordered tails of beta-catenin
Zhao B and Xue B
BMC Genomics, 17(5), 484-484 (2016)
Alma Kuechler et al.
Human genetics, 134(1), 97-109 (2014-10-20)
Recently, de novo heterozygous loss-of-function mutations in beta-catenin (CTNNB1) were described for the first time in four individuals with intellectual disability (ID), microcephaly, limited speech and (progressive) spasticity, and functional consequences of CTNNB1 deficiency were characterized in a mouse model.
Shirin Bahmanyar et al.
Genes & development, 22(1), 91-105 (2007-12-19)
beta-Catenin plays important roles in cell adhesion and gene transcription, and has been shown recently to be essential for the establishment of a bipolar mitotic spindle. Here we show that beta-catenin is a component of interphase centrosomes and that stabilization
Yang Wan et al.
Human pathology, 45(7), 1459-1465 (2014-04-22)
Osteoblastoma (OB) and osteosarcoma (OS) are 2 bone tumors that predominantly affect young adults. The clinical management of OS differs significantly from that of OB, and thus, accurate diagnosis of OB and OS is critical in determining appropriate treatment modality.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service