Skip to Content
Merck
All Photos(2)

Key Documents

C7629

Sigma-Aldrich

L-Citrulline

≥98% (TLC)

Synonym(s):

(S)-2-Amino-5-ureidopentanoic acid, L-2-Amino-5-ureidovaleric acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13N3O3
CAS Number:
Molecular Weight:
175.19
Beilstein:
1725416
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.32

Product Name

L-Citrulline, ≥98% (TLC)

Quality Level

Assay

≥98% (TLC)

form

powder

color

white to off-white

application(s)

detection

SMILES string

N[C@@H](CCCNC(N)=O)C(O)=O

InChI

1S/C6H13N3O3/c7-4(5(10)11)2-1-3-9-6(8)12/h4H,1-3,7H2,(H,10,11)(H3,8,9,12)/t4-/m0/s1

InChI key

RHGKLRLOHDJJDR-BYPYZUCNSA-N

Looking for similar products? Visit Product Comparison Guide

Application

L-Citrulline has been used in ultra-performance liquid chromatography to validate, metabolites involved in the coordination of metamorphic processes.

Biochem/physiol Actions

Intermediate in the production of nitric oxide from L-arginine by NO synthetase.
L-Citrulline, when orally administered is found to ameliorate the condition of sickle cell disease in humans. L-citrulline can be a replacement for L-arginine administration in case of defective NO synthetase. L-Citrulline supplementation is found to fulfill the purpose of NO-dependent signaling.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pharmacokinetic and pharmacodynamic properties of oral L-citrulline and L-arginine: impact on nitric oxide metabolism.
Schwedhelm E
British Journal of Clinical Pharmacology, 65(1), 51-59 (2008)
l-Citrulline and l-arginine supplementation retards the progression of high-cholesterol-diet-induced atherosclerosis in rabbits.
Hayashi T
Proceedings of the National Academy of Sciences of the USA null
Nu-Hydroxy-L-arginine Is an Intermediate in the Biosynthesis of Nitric Oxide from L-Arginin
Dennis J. Stuehr
The Journal of Biological Chemistry, 266, 6259-6263 (1991)
Metabolomic insights into system-wide coordination of vertebrate metamorphosis.
Ichu TA
BMC Developmental Biology, 14:5, 1-24 (2014)
Stephen W Scally et al.
The Journal of experimental medicine, 210(12), 2569-2582 (2013-11-06)
Rheumatoid arthritis (RA) is strongly associated with the human leukocyte antigen (HLA)-DRB1 locus that possesses the shared susceptibility epitope (SE) and the citrullination of self-antigens. We show how citrullinated aggrecan and vimentin epitopes bind to HLA-DRB1*04:01/04. Citrulline was accommodated within

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service