Skip to Content
Merck
All Photos(1)

Key Documents

C7249

Sigma-Aldrich

CI-1033

≥98% (HPLC)

Synonym(s):

Canertinib dihydrochloride, N-[4-(3-Chloro-4-fluorophenylamino)-7-(3-morpholin-4-ylpropoxy)quinazolin-6-yl]acrylamide dihydrochloride, N-[4-[(3-Chloro-4-fluorophenyl)amino]-7-[3-(4-morpholinyl)propoxy]-6-quinazolinyl]-2-propenamide dihydrochloride, PD183805

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H25ClFN5O3· 2HCl
CAS Number:
Molecular Weight:
558.86
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

solubility

DMSO: >10 mg/mL
H2O: >10 mg/mL

storage temp.

−20°C

SMILES string

Cl.Cl.Fc1ccc(Nc2ncnc3cc(OCCCN4CCOCC4)c(NC(=O)C=C)cc23)cc1Cl

InChI

1S/C24H25ClFN5O3.2ClH/c1-2-23(32)30-21-13-17-20(14-22(21)34-9-3-6-31-7-10-33-11-8-31)27-15-28-24(17)29-16-4-5-19(26)18(25)12-16;;/h2,4-5,12-15H,1,3,6-11H2,(H,30,32)(H,27,28,29);2*1H

InChI key

JZZFDCXSFTVOJY-UHFFFAOYSA-N

Gene Information

Biochem/physiol Actions

CI-1033 is a potent, irreversible ATP binding site–directed pan-ErbB tyrosine kinase inhibitor with IC50 in the low nanomolar range for EGFR, HER2, and ErbB-4.

Features and Benefits

This compound is a featured product for Kinase Phosphatase Biology research. Click here to discover more featured Kinase Phosphatase Biology products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the EGFR page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Samiksha Katiyar et al.
PloS one, 8(2), e56150-e56150 (2013-02-26)
Human African trypanosomiasis is caused by the eukaryotic microbe Trypanosoma brucei. To discover new drugs against the disease, one may use drugs in the clinic for other indications whose chemical scaffolds can be optimized via a medicinal chemistry campaign to
Alberto Ocaña et al.
Cancer treatment reviews, 35(8), 685-691 (2009-09-08)
Aberrant activation of HER2 through overexpression has been shown to play an important role in some breast cancers. Therapies against this receptor including the monoclonal antibody, trastuzumab, or the small tyrosine kinase inhibitor, lapatinib have shown to improve the prognosis
M Sánchez-Martín et al.
International journal of cancer, 131(1), 244-252 (2011-08-10)
Deregulation of ErbB/HER receptor tyrosine kinases has been linked to several types of cancer. The mechanism of activation of these receptors includes establishment of receptor dimers. Here, we have analyzed the action of different small molecule HER tyrosine kinase inhibitors
Cecilia Trinks et al.
Biochemical and biophysical research communications, 393(1), 6-10 (2010-01-26)
Epidermal growth factor (EGF) receptor tyrosine kinase inhibitors have recently been shown to display anti-neoplastic effects in human malignant myeloid cells. Our study was initiated in order to determine the effect of the pan-ErbB receptor tyrosine kinase inhibitor, canertinib (CI-1033)
Chandra P Belani
Cancer investigation, 28(4), 413-423 (2010-03-24)
Reversible epidermal growth factor receptor tyrosine kinase inhibitors are often used for the treatment of non-small cell lung cancer following failure of cytotoxic chemotherapy. While these agents are active in a subset of patients, most develop resistance and progress within

Articles

Discover Bioactive Small Molecules for Kinase Phosphatase Biology

Discover Bioactive Small Molecules for Kinase Phosphatase Biology

Discover Bioactive Small Molecules for Kinase Phosphatase Biology

Discover Bioactive Small Molecules for Kinase Phosphatase Biology

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service