Skip to Content
Merck
All Photos(1)

Documents

SML1083

Sigma-Aldrich

Ethacrynic acid

≥97% (HPLC)

Synonym(s):

2-[2,3-Dichloro-4-(2-methylene-1-oxobutyl)phenoxy]acetic acid, ECA, Etacrynic acid, [2,3-Dichloro-4-(2-ethylacryloyl)phenoxy]acetic acid, (2,3-Dichloro-4-[2-methylenebutyryl]phenoxy)acetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H12Cl2O4
CAS Number:
Molecular Weight:
303.14
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.77

Assay

≥97% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

ClC1=C(Cl)C(C(C(CC)=C)=O)=CC=C1OCC(O)=O

InChI

1S/C13H12Cl2O4/c1-3-7(2)13(18)8-4-5-9(12(15)11(8)14)19-6-10(16)17/h4-5H,2-3,6H2,1H3,(H,16,17)

InChI key

AVOLMBLBETYQHX-UHFFFAOYSA-N

Gene Information

human ... SLC12A1(6557)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Ethacrynic acid increases the cytotoxicity of chlorambucil in rat and human tumor cell lines.
Ethacrynic acid is non sulfonamide loop diuretic that is used to treat high blood pressure and the swelling caused by diseases like congestive heart failure. Ethacrynic acid blocks sodium-potassium-chloride cotransport. Also, Ethacrynic acid potently inhibits glutathione S-transferase family members. Studies show that ethacrynic acid potently inhibits Tgase-2 (transglutaminase-2) dependent metastasis of cancer cells including lung and pancreatic cancers.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Röth et al.
Experimental and clinical cardiology, 16(3), 92-96 (2011-11-09)
Oxidative stress and ischemia-reperfusion (I/R) injury are crucial in the pathogenesis of cardiovascular diseases. The antioxidant glutathione S-transferase (GST) is responsible for the high-capacity metabolic inactivation of electrophilic compounds and toxic substrates. The main objective of the present study was
Maria Font Farre et al.
Plant & cell physiology, 65(1), 128-141 (2023-11-04)
Glutathione transferases (GSTs) represent a large and diverse enzyme family involved in the detoxification of small molecules by glutathione conjugation in crops, weeds and model plants. In this study, we introduce an easy and quick assay for photoaffinity labeling of
Inhibition of rat and human glutathione S-transferase isoenzymes by ethacrynic acid and its glutathione conjugate.
Ploemen J H, et al.
Biochemical Pharmacology, 40(7), 1631-1635 (1990)
Piriyaporn Thiendedsakul et al.
Veterinary world, 15(1), 46-54 (2022-04-05)
The crocodile is a model for studying relevant sources of environmental contamination. They were determined an appropriate biomonitoring species for various toxins. The cytosolic and microsomal fraction of crocodiles plays a role in detoxifying xenobiotics. Cytochrome P450 1A2 (CYP1A2) metabolizes
Ruina Wang et al.
Nature communications, 14(1), 5984-5984 (2023-09-27)
Induction of hypothermia during hibernation/torpor enables certain mammals to survive under extreme environmental conditions. However, pharmacological induction of hypothermia in most mammals remains a huge challenge. Here we show that a natural product P57 promptly induces hypothermia and decreases energy

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service