Skip to Content
Merck
All Photos(1)

Documents

C6788

Sigma-Aldrich

4-Chloro-1-naphthol

peroxidase substrate, tablet, suitable for use in immunoblotting procedures

Synonym(s):

4-CN, 4-chloronaphthalen-1-ol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC10H6OH
CAS Number:
Molecular Weight:
178.61
Beilstein:
509750
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

product name

4-Chloro-1-naphthol, tablet

form

tablet

mp

118-121 °C (lit.)

solubility

methanol: 10 mL, clear, colorless

storage temp.

−20°C

SMILES string

Oc1ccc(Cl)c2ccccc12

InChI

1S/C10H7ClO/c11-9-5-6-10(12)8-4-2-1-3-7(8)9/h1-6,12H

InChI key

LVSPDZAGCBEQAV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Specificity

4-Chloro-1-Naphthol is a peroxidase substrate suitable for use in immunoblotting procedures. This substrate produces an insoluble end product that is blue in color and can be observed visually.

Application

4-Chloro-1-naphthol has been used as a substrate in enzyme-linked immunospot (ELISpot) assay for detection of the spots. It has also been used as a precipitating chromogenic substrate for visualizing the antigen−antibody-reactive bands following western blotting.

Quantity

Contains 30 mg substrate per tablet.

Reconstitution

Dissolve 1 tablet in 10 ml of methanol. Add 2 ml of methanol stock solution to 10 ml of triethanolamine buffered saline, pH 7.5. Add 5 μl of fresh 30% hydrogen peroxide (Sigma Product No. H-1009) immediately prior to use.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Ellakany et al.
Avian pathology : journal of the W.V.P.A, 27(6), 547-554 (2008-05-17)
Two experiments were conducted to test the sensitivity of Western blotting for detection of M. gallisepticum antibodies in respiratory washings and sera of infected chickens by mouse monoclonal antibodies to chicken IgG, IgM and IgA. In the first experiment, birds
Daniela Staubli et al.
Parasitology research, 99(6), 648-658 (2006-05-24)
Neospora caninum ranges among the major causes of infectious abortion in cattle worldwide. The present study was designed to improve the serodiagnostic tools by complementing a conventional ELISA with a highly sensitive and species-specific N. caninum immunoblot. To evaluate this
Precolostral serology in calves born from Neospora-seropositive mothers
Staubli D, et al.
Parasitology Research, 99(4), 398-404 (2006)
Karl Albert Brokstad et al.
The Journal of infectious diseases, 185(7), 878-884 (2002-03-29)
The immune response in the nasal mucosa to influenza vaccination in 23 patients scheduled for tonsillectomy was studied. A statistically significant increase in influenza virus-specific serum and oral fluid antibodies was observed 7 days after vaccination. The numbers of influenza
B Wróblewska et al.
Food & function, 9(3), 1500-1509 (2018-02-09)
The objective of this work was to identify the nutritional and physiological effects of commercial soy and whey protein preparations. Wistar rats were fed with soy (S), whey (W), or casein (C) preparations as the sole dietary protein source. The

Articles

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service