Skip to Content
Merck
All Photos(1)

Key Documents

C222

Sigma-Aldrich

Cathine hydrochloride

≥97.5% (HPLC), powder, anorexic

Synonym(s):

threo-2-Amino-1-hydroxy-1-phenyl propane hydrochloride, Pseudonorephedrine hydrochloride, S,S-Norpseudoephedrine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H13NO · HCl
CAS Number:
Molecular Weight:
187.67
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

Cathine hydrochloride,

form

powder

Quality Level

drug control

USDEA Schedule IV; Home Office Schedule 3; psychotrope (France); kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada; psicótropo (Spain); Decreto Lei 15/93: Tabela IIB (Portugal)

storage condition

desiccated

color

white to off-white

mp

172-175 °C

solubility

H2O: 15 mg/mL, clear
0.1 M HCl: soluble
ethanol: soluble

application(s)

forensics and toxicology

SMILES string

Cl[H].C[C@H](N)[C@@H](O)c1ccccc1

InChI

1S/C9H13NO.ClH/c1-7(10)9(11)8-5-3-2-4-6-8;/h2-7,9,11H,10H2,1H3;1H/t7-,9+;/m0./s1

InChI key

DYWNLSQWJMTVGJ-DKXTVVGFSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Anorexic; with pharmacological effects similar to those of the amphetamines.

Caution

Photosensitive

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E A Pehek et al.
Pharmacology, biochemistry, and behavior, 36(2), 267-271 (1990-06-01)
The effects of the psychostimulant (+)cathine (norpseudoephedrine) were examined in a two-choice, food-motivated, drug-discrimination paradigm. Rats were able to discriminate cathine from vehicle and this effect was dose- and time-dependent. Prior administration of cathine resulted in a diminished response (tolerance)
M B Ahmed et al.
Journal of ethnopharmacology, 39(3), 213-216 (1993-08-01)
Catha edulis is a plant that grows in certain areas of East Africa and the Arab Peninsula. The stimulating properties of fresh material were described more than seven centuries ago and today the habit of inducing a state of euphoria
John S Chappell et al.
Forensic science international, 195(1-3), 108-120 (2009-12-29)
A primary concern with the forensic analysis of the khat plant (Catha edulis) has been the need to preserve the principle psychoactive component, cathinone, which converts to the less-active substance, cathine, after harvesting. The loss of cathinone has serious legal
Hyeon-Kyu Lee et al.
The Journal of organic chemistry, 77(12), 5454-5460 (2012-05-25)
Each of the enantiomers of both norephedrine and norpseudoephedrine were stereoselectively prepared from the common, prochiral cyclic sulfamidate imine of racemic 1-hydroxy-1-phenyl-propan-2-one by employing asymmetric transfer hydrogenation (ATH) catalyzed by the well-defined chiral Rh-complexes, (S,S)- or (R,R)-Cp*RhCl(TsDPEN), and HCO(2)H/Et(3)N as
Richard Hoffman et al.
Journal of ethnopharmacology, 132(3), 554-563 (2010-06-18)
Although there is a rich body of research available regarding the effect of acute and chronic khat dosing in animal models, research on the behavioral and cognitive effects of khat in human subjects is not extensive and several of the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service