Skip to Content
Merck

Skip To

C5499

Cytochrome c Oxidase from bovine heart

5 mg protein/mL

greener alternative

Greener Alternative

Synonym(s):

EC 1.9.3.1, Ferrocytochrome-c:oxygen oxidoreductase, Complex IV

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352204
NACRES:
NA.32
EC Number:
232-586-0
MDL number:
EC Number:
Specific activity:
≥20 units/mg protein
Biological source:
bovine heart
Concentration:
5 mg protein/mL
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

bovine heart

Quality Segment

form

buffered aqueous solution

specific activity

≥20 units/mg protein

mol wt

300-350 kDa by gel filtration

greener alternative product characteristics

Waste Prevention
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

5 mg protein/mL

greener alternative category

shipped in

dry ice

storage temp.

−20°C

General description

Research area: Cell cycle

Cytochrome c oxidase contains 13 subunits, encoded by mitochondrial genes and nuclear genome. It is comprised of two heme groups (Fea and Fea3) and two copper centers, CuA and CuB.

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency and waste prevention when used in biofuel cell research. For more information see the article in biofiles.

Application

Cytochrome c Oxidase (CcO) from bovine heart has been used:
  • to study the effect of oxidized glutathione (GSSG) on the number of total free cysteines
  • in a histochemical assay to measure CcO activity in tissue sections
  • to study the redox reactions of cytochrome c in isolated mitochondria exposed to blue or red lasers using resonance Raman spectroscopy

Biochem/physiol Actions

Cytochrome c oxidase is the principal terminal oxidase of high oxygen affinity in the aerobic metabolism of all animals, plants, yeasts and some bacteria.
Cytochrome c oxidase is the principal terminal oxidase of high oxygen affinity in the aerobic metabolism of all animals, plants, yeasts and some bacteria. It is present in the mitochondria of the more highly developed cells and in the cytoplasmic membrane of bacteria. Cytochrome c oxidase catalyses the electron transfer from cytochrome c to O2. This electron-transfer process produces a proton gradient across the membrane, which in turn drives the production of ATP. This enzyme is unique in providing energy for the cell bycoupling the electron transport through the cytochrome chain with the process of oxidative phosphorylation.

Physical form

Supplied as a solution in 25 mM Tris-HCl buffer, pH 7.8, 5 mM EDTA, and 39 mM n-dodecyl β·-D-maltoside.

Other Notes

One unit of cytochrome c oxidase will oxidize 1.0 μmole of ferrocytochrome c per min. at 25 °C at pH 6.0.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
P6621C3131P7633
description

5 mg protein/mL

description

≥200 units/mg protein

description

≥95% based on Mol. Wt. 12,327 basis, powder, suitable for mammalian cell culture

description

Type I, lyophilized powder, 10-50 units/mg protein

specific activity

≥20 units/mg protein

specific activity

≥200 units/mg protein

specific activity

-

specific activity

10-50 units/mg protein

biological source

bovine heart

biological source

-

biological source

bovine heart

biological source

bacterial (Clostridium perfringens)

concentration

5 mg protein/mL

concentration

-

concentration

-

concentration

-

form

buffered aqueous solution

form

lyophilized powder

form

powder

form

lyophilized powder

shipped in

dry ice

shipped in

-

shipped in

-

shipped in

-

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C


Storage Class

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters